GVS Buffer Swabs
Easily collect microbial surface samples for analysis
  • Excellent for flat or convoluted surfaces
  • Does not contain bacteriostatic or bactericidal compounds
  • Sterile packed and ready-for-use
Use these cotton swabs for collecting surface contamination from flat or convoluted surfaces for analysis. Swabs do not contain any bactericidal compounds ensuring accurate results. After collecting the samples, simply transport to a laboratory for culture and enumeration. Each self-standing, round-bottomed plastic vial contains clear buffer with a cotton swab secured inside the screw cap. Store vials in temperatures between 2 and 8°C (35.6 and 46.4°F).
£642.97 - £2,020.15GBP / Pkg of 125
£771.56 - £2,424.18GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Swab Total Length (InchShort) Description
Availability Pricing
GVS Buffer Swabs, 4 mL; 125/PK 7890942 Mfr # 10498305
COMPARE
Cotton swab, polypropylene tube, and polypropylene cap with silicone washer 3 5/64 Buffer Swabs, 4 mL; 125/PK
£642.97GBP / Pkg of 125
£771.56GBP (incl VAT)
GVS Buffer Swabs, 4 mL; 500/PK 7890943 Mfr # 10498306
COMPARE
Cotton swab, polypropylene tube, and polypropylene cap with silicone washer 3 5/64 Buffer Swabs, 4 mL; 500/PK
£2,020.15GBP / Pkg of 500
£2,424.18GBP (incl VAT)