Green Polyester Adhesives Tape
Green polyester tape is ideal for high-temperature masking applications in most powder coat painting, anodizing, and plating situationsan economical alternative to polymide tape. Tape is also excellent for use as die-cut material on liner. Withstands temperatures as high as 310°F (205°C) and removes cleanly from nearly all surfaces. This tape is resistant to corrosive chemicals, tearing, and breaking. Come on a 3" diameter core.
£25.72GBP / Roll of 1
£30.86GBP (incl VAT)
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (ft) Material Thickness (mil)
Availability Pricing
COMPARE
216 Polyester 1.5
£25.72GBP / Roll of 1
£30.86GBP (incl VAT)