Glass Tempering Beaker
Easily temper hot or cold fluids
  • Double walled glass beaker
  • Includes inlet and outlet ports for controlling temperature
Constructed of durable borosilicate glass, these double-walled beakers are ideal for tempering hot and cold liquids. Inlet and outlet ports provide easy connection to a circulator to maintain constant sample temperatures.
£248.89 - £316.09GBP / Each
£298.67 - £379.31GBP (incl VAT)
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Material
Availability Pricing
COMPARE
250 Borosilicate Glass
£248.89GBP / Each
£298.67GBP (incl VAT)
COMPARE
600 Borosilicate Glass
£267.98GBP / Each
£321.58GBP (incl VAT)
COMPARE
2000 Borosilicate Glass
£316.09GBP / Each
£379.31GBP (incl VAT)