Gems Sensors & Controls Liquid Flow Rate Indicators
High visibility rotor and pulsed output provide accurate flow rate output with visual flow verifications
  • Hall-effect sensor produces a 4.5 to 24 VDC pulse output proportional to the flow
  • Either port may be used for incoming flow
  • Removable lens for quick and easy cleaning
These flow indicators combine a high visibility rotor with solid state electronics in a compact body to provide accurate flow indication. Each of the six-rotor arm features a magnet that excites the Hall-effect sensor in the indicator body, producing a 4.5 to 24 VDC pulse output. Frequency of the output pulse is proportional to the flow rate and ranges from approximately 15 Hz at low flow to 225 Hz at high flow. Send the signals to a digital rate meter to view the flow rate. Polypropylene body and PPS composite rotor construction.
£215.90GBP / Each
£259.08GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Flow Rate (GPM) Min Flow Rate (GPM) Process Connection
Availability Pricing
Gems Sensors & Controls Flow Rate Indicator, PP, 1/4" NPT(F), 0.1 to 5 GPM 4852702 Mfr # 155421
COMPARE
5 0.1 1/4" NPT(F)
£215.90GBP / Each
£259.08GBP (incl VAT)
Gems Sensors & Controls Flow Rate Indicator, PP, 1/2" NPT(F), 1.5 to 20 GPM 4852703 Mfr # 155481
COMPARE
20 1.5 1/2" NPT(F)
£215.90GBP / Each
£259.08GBP (incl VAT)