Gems Sensors & Controls Liquid Flow Indicators
Bright orange rotor provides visual flow verifications
  • Rotor spins at a rate proportional to the flow
  • Either port may be used for incoming flow
  • Removable lens for quick and easy cleaning
Estimate the flow rate or confirm flow simply by viewing the speed of the turning rotor. Easily view a bright orange rotor that spins at a rate proportional to the flow through a clear polysulfone lens. For best results, use indicator with a 150 µm filter on the inlet and fluids with a viscosity less than 34 centipoise. Polypropylene body and molded nylon rotor construction.
£104.55GBP / Each
£125.46GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Flow Rate (GPM) Min Flow Rate (GPM) Process Connection
Availability Pricing
Gems Sensors & Controls Flow Indicator, PP, 1/2" NPT(F), 1.5 to 20 GPM 4852700 Mfr # 155480
COMPARE
20 1.5 1/2" NPT(F)
£104.55GBP / Each
£125.46GBP (incl VAT)
Gems Sensors & Controls Flow Indicator, PP, 1/4" NPT(F), 0.1 to 5 GPM 4852701 Mfr # 155420
COMPARE
5 0.1 1/4" NPT(F)
£104.55GBP / Each
£125.46GBP (incl VAT)