Extech® Waterproof Pocket Stem Thermometers
Pocket thermometer with stainless steel stem
  • 2.75'' (70mm) Stainless steel stem
  • Temperature: -40 to 392°F (-40 to 200°C)
  • MAX/MIN recall of high and low temperatures
  • Data Hold and Auto power Off features
  • ±2°F/1°C accuracy, 0.1°/1° resolution
Reduced Tip Waterproof Thermometer with antimicrobial protection, has a 3'' long stem and a 1.5mm diameter tip. Features °F/°C switchable ranges, a hold function, a display that updates every 1.5 seconds and low battery indicator. Use ON/OFF button to conserve battery life or the unit will automatically shut off after 35 minutes..
£24.89GBP / Each
£29.87GBP (incl VAT)
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Min Temperature (° F) Stem Length (in)
Availability Pricing
Extech Digital Waterproof Pocket Stem Thermometer 8648723 Mfr # 39240
COMPARE
392 -40 2 3/4
£24.89GBP / Each
£29.87GBP (incl VAT)