Extech Type-K Thermocouple Pipe Clamp Probes
Easily measure pipe temperature
  • Works on pipes from 6.4 to 38 mm (1/4 to 1-1/2") in diameter
The spring-loaded jaw securely clamps on pipes for hands-free temperature measurement. Compatible with most multimeters and thermometers that have Type K temperature function.
£43.15 - £82.97GBP / Each
£51.78 - £99.56GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° C)
Availability Pricing
Extech TP200 Type-K, Pipe Clamp Probe with Spring-Loaded Jaw, Mini-Connector, 1/4" to 1-1/2" 9500126 Mfr # TP200
COMPARE
93
£43.15GBP / Each
£51.78GBP (incl VAT)
Extech TP400 Type-K, High-Temperature Pipe Clamp Probe with Spring-Loaded Jaw, Mini-Connector, 1/4" to 1-1/2" 9500127 Mfr # TP400
COMPARE
232
£82.97GBP / Each
£99.56GBP (incl VAT)