Extech Dual Laser Infrared Thermometers
Dual-laser targeting indicates the ideal measuring distance to ensure accuracy
  • Fast 0.15-second response time
  • Rugged, double-molded housing
These infrared thermometers features dual lasers that converge to a 1" target spot to ensure accurate targeting and temperature measurement. All thermometers feature a backlit dual display for clear and easy reading. You can program the adjustable high/low setpoints with audible alarm to alert you when set point temperature is exceeded. Adjustable emissivity for increased measurement accuracy on virtually any surface color, finish, or texture. Other features include Fahrenheit or Celsius selectable units, current and maximum readings, auto hold, continuous scanning, and auto power off.
.
£198.29GBP / Each
£237.95GBP (incl VAT)
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Distance To Target Size Ratio (D:S) Emissivity Max Temperature (° F)
Availability Pricing
Extech 42512 Dual Laser Infrared Thermometer (30: 1) 9044064 Mfr # 42512
COMPARE
30:1 Adjustable 1832
£198.29GBP / Each
£237.95GBP (incl VAT)