Excelon Platinum-Cured Silicone Tubing
Pumpable tubing that is resilient across extreme temperatures (-60°C to 200°C) and FDA compliant, making it a perfect fit for a diverse range of applications
  • Suitable for food, beverage, laboratory, and more
  • Enhanced durability offers superior resilience and stretchability to minimize replacements
  • The natural translucent color provides visual assurance
  • Withstands repeated sterilization, making it ideal for frequent cycles
  • Free of BPA, latex, and phthalates
  • Odorless, tasteless, and inert
Platinum-cured silicone tubing offers excellent reliability, purity, and versatility. Engineered with precision and crafted from FDA-compliant materials, this tubing ensures the safe transfer of fluids across a multitude of industries, including pharmaceuticals, food and beverage, laboratory, and more. Its unique composition allows it to withstand extreme temperatures while maintaining consistent performance, providing assurance in even the most demanding environments. The tubing's natural translucent color enables visual confirmation of fluid flow, ensuring integrity throughout processing. Additionally, its superior durability and resistance to compression set minimize the need for frequent replacements.
£31.50 - £453.34GBP / Pkg of 1
£37.80 - £544.01GBP (incl VAT)
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubing ID (in) Tubing OD (in) Wall Thickness (in)
Availability Pricing
Excelon Platinum-Cured Silicone Tubing, 70 Shore A, 1/16" ID x 1/8" OD; 50 Ft 5011784 Mfr # 28901705
COMPARE
1/16 1/8 1/32
£31.50GBP / Pkg of 1
£37.80GBP (incl VAT)
Excelon Platinum-Cured Silicone Tubing, 70 Shore A, 1/8" ID x 1/4" OD; 50 Ft 5011785 Mfr # 280205
COMPARE
1/8 1/4 1/16
£57.56GBP / Pkg of 1
£69.07GBP (incl VAT)
Excelon Platinum-Cured Silicone Tubing, 70 Shore A, 3/16" ID x 5/16" OD; 50 Ft 5011786 Mfr # 280305
COMPARE
3/16 5/16 1/16
£84.27GBP / Pkg of 1
£101.12GBP (incl VAT)
Excelon Platinum-Cured Silicone Tubing, 70 Shore A, 3/16" ID x 3/8" OD; 50 Ft 5011787 Mfr # 280405
COMPARE
3/16 3/8 3/32
£136.50GBP / Pkg of 1
£163.80GBP (incl VAT)
Excelon Platinum-Cured Silicone Tubing, 70 Shore A, 1/4" ID x 3/8" OD; 50 Ft 5011788 Mfr # 280605
COMPARE
1/4 3/8 1/16
£95.38GBP / Pkg of 1
£114.46GBP (incl VAT)
Excelon Platinum-Cured Silicone Tubing, 70 Shore A, 1/4" ID x 7/16" OD; 50 Ft 5011789 Mfr # 280705
COMPARE
1/4 7/16 3/32
£158.71GBP / Pkg of 1
£190.45GBP (incl VAT)
Excelon Platinum-Cured Silicone Tubing, 70 Shore A, 1/4" ID x 1/2" OD; 50 Ft 5011790 Mfr # 280805
COMPARE
1/4 1/2 1/8
£228.89GBP / Pkg of 1
£274.67GBP (incl VAT)
Excelon Platinum-Cured Silicone Tubing, 70 Shore A, 5/16" ID x 1/2" OD; 50 Ft 5011791 Mfr # 281005
COMPARE
5/16 1/2 3/32
£186.30GBP / Pkg of 1
£223.56GBP (incl VAT)
Excelon Platinum-Cured Silicone Tubing, 70 Shore A, 3/8" ID x 1/2" OD; 50 Ft 5011792 Mfr # 281205
COMPARE
3/8 1/2 1/16
£133.52GBP / Pkg of 1
£160.22GBP (incl VAT)
Excelon Platinum-Cured Silicone Tubing, 70 Shore A, 3/8" ID x 9/16" OD; 50 Ft 5011793 Mfr # 281305
COMPARE
3/8 9/16 3/32
£215.19GBP / Pkg of 1
£258.23GBP (incl VAT)
Excelon Platinum-Cured Silicone Tubing, 70 Shore A, 3/8" ID x 5/8" OD; 50 Ft 5011794 Mfr # 281405
COMPARE
3/8 5/8 1/8
£305.93GBP / Pkg of 1
£367.12GBP (incl VAT)
Excelon Platinum-Cured Silicone Tubing, 70 Shore A, 1/2" ID x 11/16" OD; 50 Ft 5011795 Mfr # 281905
COMPARE
1/2 11/16 3/32
£273.71GBP / Pkg of 1
£328.45GBP (incl VAT)
Excelon Platinum-Cured Silicone Tubing, 70 Shore A, 1/2" ID x 3/4" OD; 50 Ft 5011796 Mfr # 282005
COMPARE
1/2 3/4 1/8
£382.23GBP / Pkg of 1
£458.68GBP (incl VAT)
Excelon Platinum-Cured Silicone Tubing, 70 Shore A, 5/8" ID x 7/8" OD; 50 Ft 5011797 Mfr # 282605
COMPARE
5/8 7/8 1/8
£453.34GBP / Pkg of 1
£544.01GBP (incl VAT)
Showing 1- 14 of 14