Excelon Platinum-Cured Braid-Reinforced Silicone Tubing
Added strength and durability while maintaining the purity and reliability expected from silicone tubing
  • Features a reinforced layer, making it ideal for high-pressure applications
  • Crafted from FDA compliant materials, it's suitable for food, beverage, laboratory, and more
  • Offers superior resilience and stretchability to minimize replacements
  • The natural translucent color provides visual assurance
  • Withstands repeated sterilization, making it ideal for frequent cycles
  • Free of BPA, latex, and phthalates
  • Odorless, tasteless, and inert
The braided version of platinum-cured silicone tubing represents an evolution in fluid transfer technology, combining the reliability and purity of silicone with enhanced durability. Constructed with a braided reinforcement layer, this tubing offers increased strength and resistance to kinking, making it ideal for applications requiring higher pressure or more rigorous handling. Like its non-braided counterpart, it maintains FDA compliance and can withstand extreme temperatures, ensuring safe fluid transfer across various industries. With its superior durability, braided silicone tubing sets a new standard in performance for fluid handling applications.
£115.19 - £731.86GBP / Pkg of 1
£138.23 - £878.23GBP (incl VAT)
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubing ID (in) Tubing OD (in) Wall Thickness (in)
Availability Pricing
Excelon Braid-Reinforced Silicone Tubing, 0.125" ID x 0.365" OD; 25 Ft 5011770 Mfr # 2836525BR
COMPARE
1/8 23/64 1/8
£115.19GBP / Pkg of 1
£138.23GBP (incl VAT)
Excelon Braid-Reinforced Silicone Tubing, 0.187" ID x 0.447" OD; 25 Ft 5011771 Mfr # 2844725BR
COMPARE
3/16 29/64 1/8
£160.00GBP / Pkg of 1
£192.00GBP (incl VAT)
Excelon Braid-Reinforced Silicone Tubing, 0.188" ID x 0.452" OD; 25 Ft 5011772 Mfr # 2846225BR
COMPARE
3/16 29/64 1/8
£173.89GBP / Pkg of 1
£208.67GBP (incl VAT)
Excelon Braid-Reinforced Silicone Tubing, 0.250" ID x 0.500" OD; 25 Ft 5011773 Mfr # 280825BR
COMPARE
1/4 1/2 1/8
£182.78GBP / Pkg of 1
£219.34GBP (incl VAT)
Excelon Braid-Reinforced Silicone Tubing, 0.250" ID x 0.520" OD; 25 Ft 5011774 Mfr # 2852025BR
COMPARE
1/4 33/64 1/8
£202.23GBP / Pkg of 1
£242.68GBP (incl VAT)
Excelon Braid-Reinforced Silicone Tubing, 0.312" ID x 0.592" OD; 25 Ft 5011775 Mfr # 2859225BR
COMPARE
5/16 19/32 9/64
£247.41GBP / Pkg of 1
£296.89GBP (incl VAT)
Excelon Braid-Reinforced Silicone Tubing, 0.375" ID x 0.625" OD; 25 Ft 5011776 Mfr # 281425BR
COMPARE
3/8 5/8 1/8
£244.08GBP / Pkg of 1
£292.90GBP (incl VAT)
Excelon Braid-Reinforced Silicone Tubing, 0.375" ID x 0.655" OD; 25 Ft 5011777 Mfr # 2865525BR
COMPARE
3/8 21/32 9/64
£280.75GBP / Pkg of 1
£336.90GBP (incl VAT)
Excelon Braid-Reinforced Silicone Tubing, 0.500" ID x 0.800" OD; 25 Ft 5011778 Mfr # 2880025BR
COMPARE
1/2 51/64 5/32
£380.00GBP / Pkg of 1
£456.00GBP (incl VAT)
Excelon Braid-Reinforced Silicone Tubing, 0.500" ID x 0.875" OD; 25 Ft 5011779 Mfr # 28S1825BR
COMPARE
1/2 7/8 3/16
£497.78GBP / Pkg of 1
£597.34GBP (incl VAT)
Excelon Braid-Reinforced Silicone Tubing, 0.625" ID x 0.965" OD; 25 Ft 5011780 Mfr # 2896525BR
COMPARE
5/8 31/32 11/64
£521.49GBP / Pkg of 1
£625.79GBP (incl VAT)
Excelon Braid-Reinforced Silicone Tubing, 0.625" ID x 0.990" OD; 25 Ft 5011781 Mfr # 2899025BR
COMPARE
5/8 63/64 11/64
£569.63GBP / Pkg of 1
£683.56GBP (incl VAT)
Excelon Braid-Reinforced Silicone Tubing, 0.750" ID x 1.100" OD; 50 Ft 5011782 Mfr # 28110025BR
COMPARE
3/4 1 1/10 11/64
£625.19GBP / Pkg of 1
£750.23GBP (incl VAT)
Excelon Braid-Reinforced Silicone Tubing, 0.875" ID x 1.235" OD; 25 Ft 5011783 Mfr # 28123525BR
COMPARE
7/8 1 15/64 11/64
£731.86GBP / Pkg of 1
£878.23GBP (incl VAT)
Showing 1- 14 of 14