Environmental Express® Vacuum Filter Funnels
Convenient units are perfect for vacuum filtration
  • Ideal for larger volume filtration
  • Comes standard with pre-inserted filters
These filters fit snugly in most standard vacuum flasks. There are preassembled and disposable options perfect for any number of applications.
£56.25 - £400.42GBP / Pkg of 1
£67.50 - £480.50GBP (incl VAT)
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Membrane Diameter (mm) Membrane Material Pore Size (µm)
Availability Pricing
COMPARE
25 Polytetrafluoroethylene (PTFE) 0.2
£56.25GBP / Pkg of 1
£67.50GBP (incl VAT)
COMPARE
25 Glass Microfiber (GMF) 0.7
£112.12GBP / Pkg of 1
£134.54GBP (incl VAT)
COMPARE
25 Mixed Cellulose Ester (MCE) 0.45
£101.64GBP / Pkg of 1
£121.97GBP (incl VAT)
COMPARE
47 Mixed Cellulose Ester (MCE) 0.45
£400.42GBP / Pkg of 80
£480.50GBP (incl VAT)
COMPARE
47 Polyethersulfone (PES) 0.45
£182.06GBP / Pkg of 50
£218.47GBP (incl VAT)