Environmental Express® TCLP Bottles
Ideal for commercial and homemade rotary extractors
  • Bottle collars or pads are necessary for most bottles to safely fit in rotary agitators
  • All bottles listed, except 35208-61 and 35208-62, have PTFE-lined caps
  • Compatible with Environmental Express heavy-duty rotator
£32.67 - £320.06GBP / Each
£39.20 - £384.07GBP (incl VAT)
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Autoclavable Capacity (L) Material
Availability Pricing
COMPARE
3 Glass
£50.90GBP / Pkg of 4
£61.08GBP (incl VAT)
COMPARE
3 Polyethylene(PE)
£32.67GBP / Pkg of 4
£39.20GBP (incl VAT)
COMPARE
2.2 Glass
£320.06GBP / Pkg of 1
£384.07GBP (incl VAT)
COMPARE
2.2 Glass
£273.05GBP / Pkg of 4
£327.66GBP (incl VAT)
COMPARE
3 Glass
£311.90GBP / Pkg of 1
£374.28GBP (incl VAT)
COMPARE
2.2 Glass
£70.55GBP / Each
£84.66GBP (incl VAT)