Environmental Express Oakton® Replacement ORP Sensors for Testrs
Replace sensor modules at the fraction of the cost of a new pocket tester -when calibration fails or gives fluctuating readings, it is time to change the sensor
  • Replaceable sensors for durability and longevity
  • Easily replace sensor module with a twist—no tools required
£73.42 - £185.81GBP / Each
£88.10 - £222.97GBP (incl VAT)
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Use With
Availability Pricing
COMPARE
35634-45
£185.81GBP / Each
£222.97GBP (incl VAT)
COMPARE
35634-54
£73.42GBP / Each
£88.10GBP (incl VAT)
COMPARE
35634-32
£87.39GBP / Each
£104.87GBP (incl VAT)