Environmental Express®­ HotBlock® 200 Digestion Heating Blocks
Same dependability with increased temperature range
  • Reach higher temperatures—perform more procedures
  • PTFE-coated blocks for chemical and corrosion resistance
  • Included polycarbonate transfer racks set on the block for easy placement and removal
Perform a wider range of experiments and digestions with the expanded temperature range—up to 200°C* with the HotBlock 200. Blocks are available with 25 to 96 wells, accepting cup sizes of 15, 50, or 100 mL, and controllers accept one or two block heaters.
Our blocks continue to be constructed of metals-free materials to avoid cross contamination and provide chemical resistance. The block housing is made of durable Kydex, and the block is constructed of PTFE-coated graphite. Polycarbonate racks allow for easy insertion and removal of multiple digestion cups.
£2,950.41 - £4,591.50GBP / Each
£3,540.49 - £5,509.80GBP (incl VAT)
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Block Number Of Openings Block - Tube / Vial Size Accepted (mL) Power (VAC)
Availability Pricing
COMPARE
96 15 240
£3,954.02GBP / Each
£4,744.82GBP (incl VAT)
COMPARE
96 15 120
£3,574.80GBP / Each
£4,289.76GBP (incl VAT)
COMPARE
36 50 240
£3,295.01GBP / Each
£3,954.01GBP (incl VAT)
COMPARE
36 50 120
£2,978.05GBP / Each
£3,573.66GBP (incl VAT)
COMPARE
54 50 240
£4,136.97GBP / Each
£4,964.36GBP (incl VAT)
COMPARE
54 50 120
£3,343.09GBP / Each
£4,011.71GBP (incl VAT)
COMPARE
25 100 240
£4,329.15GBP / Each
£5,194.98GBP (incl VAT)
COMPARE
25 100 120
£2,950.41GBP / Each
£3,540.49GBP (incl VAT)
COMPARE
35 100 240
£4,591.50GBP / Each
£5,509.80GBP (incl VAT)
COMPARE
35 100 120
£3,343.09GBP / Each
£4,011.71GBP (incl VAT)
Showing 1- 10 of 10