Environmental Express® CertiTube™ Autosampler Tubes and Vials
ICP MS/OES analysis vials that fit common industry autosampler racks for metals analysis
  • Excellent chemical resistance for a wide range of applications
  • Fits common 60-place rack autosamplers for ICP-OES and ICP-MS
  • Low metals certification for interference-free results
  • Eliminate glassware and dilute directly in tube with Class A tolerances
  • Reduce error by eliminating transfer steps
Environmental Express® self-manufactured autosampler tubes and vials are essential for accelerating lab procedures and obtaining reliable analytical results. Made from clarified homopolymer polypropylene, they offer excellent chemical resistance to aggressive metals digestion acids. CertiTubes™ are certified for low metals background and manufactured to Class A volume tolerances allowing for direct dilution in the tubes, eliminating the need for glassware and reducing prep errors. Designed for use with standard industry 60-place rack autosamplers for ICP-OES and ICP-MS instrumentation they are easily integrated into existing laboratory SOPs.
£158.72 - £167.94GBP / Pkg of 2000
£190.46 - £201.53GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Height (mm) Outside Diameter (mm) Volume (mL)
Availability Pricing
COMPARE
100 16 14
£167.94GBP / Pkg of 2000
£201.53GBP (incl VAT)
COMPARE
100 17 16.5
£158.72GBP / Pkg of 2000
£190.46GBP (incl VAT)