Eldon James SeriesLock® Quick-Disconnect Fittings, Large Hose Barb & Sanitary Flange Insert, Kynar® PVDF
Specially designed quick-disconnect fitting provides a high-volume flow at low line pressures
  • Spring-free flow path® design
  • Audible click ensures secure connection
  • Safety slide release minimizes accidental disconnects
  • Sterile quick disconnect
  • Make quick and easy sanitary connections
  • High flow yet compact to fit in tight spaces
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
The SeriesLock® quick disconnect fittings with spring-free fluid path allow a high-volume flow while minimizing turbulence, foaming, and bubbles. The SeriesLock® quick disconnect fittings with spring-free fluid path allow a high-volume flow while minimizing turbulence, foaming, and bubbles. For a secure, leak-free connection, push a large fitting insert into a large fitting body until you hear an audible “click.” Simply use the safety slide release to disconnect the fittings. The slide release eliminates accidental separation while offering easy disconnect with one hand even while wearing gloves. Recessed connection features protect contact surfaces and prolong the life of the O-rings.
Kynar PVDF quick-disconnect fittings are biocompatible and exhibit excellent UV, chemical, corrosion, and abrasion resistance. Transparent fittings allow easy viewing of fluid flow. Use fittings in temperatures ranging from 0 to 121°C (32 to 250°F).
£5.26 - £28.52GBP / Each
£6.31 - £34.22GBP (incl VAT)
30 variations of this product are available.
See All Product Options Below
30 PRODUCT OPTIONS
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Connection Type Valve
Availability Pricing
Eldon James SeriesLock® Quick-Disconnect Fitting, Hose Barb Insert, Kynar PVDF, Valved, Viton O-Ring, SS Spring, Large, 1/2" ID 5013866 Mfr # B-MV-K-VS-8
COMPARE
1/2 Quick-Disconnect Insert x Hose Barb Valved
£15.86GBP / Each
£19.03GBP (incl VAT)
Eldon James SeriesLock® Quick-Disconnect Fitting, Hose Barb Insert, Kynar PVDF, Valved, Viton O-Ring, SS Heavy Spring, Large, 1/2" ID 5013867 Mfr # B-MV-K-VS-8H
COMPARE
1/2 Quick-Disconnect Insert x Hose Barb, Heavy Spring Valved
£22.00GBP / Each
£26.40GBP (incl VAT)
Eldon James SeriesLock® Quick-Disconnect Fitting, Hose Barb Insert, Kynar PVDF, Valved, Viton O-Ring, SS Spring, Large, 5/8" ID 5013868 Mfr # B-MV-K-VS-10
COMPARE
5/8 Quick-Disconnect Insert x Hose Barb Valved
£15.86GBP / Each
£19.03GBP (incl VAT)
Eldon James SeriesLock® Quick-Disconnect Fitting, Hose Barb Insert, Kynar PVDF, Valved, Viton O-Ring, SS Heavy Spring, Large, 5/8" ID 5013869 Mfr # B-MV-K-VS-10H
COMPARE
5/8 Quick-Disconnect Insert x Hose Barb, Heavy Spring Valved
£22.38GBP / Each
£26.86GBP (incl VAT)
Eldon James SeriesLock® Quick-Disconnect Fitting, Hose Barb Insert, Kynar PVDF, Valved, Viton O-Ring, SS Spring, Large, 3/4" ID 5013870 Mfr # B-MV-K-VS-12
COMPARE
3/4 Quick-Disconnect Insert x Hose Barb Valved
£16.23GBP / Each
£19.48GBP (incl VAT)
Eldon James SeriesLock® Quick-Disconnect Fitting, Hose Barb Insert, Kynar PVDF, Valved, Viton O-Ring, SS Heavy Spring, Large, 3/4" ID 5013871 Mfr # B-MV-K-VS-12H
COMPARE
3/4 Valved
£22.38GBP / Each
£26.86GBP (incl VAT)
Eldon James SeriesLock® Quick-Disconnect Fitting, Hose Barb Insert, Kynar PVDF, Non-Valved, Large, 1/2" ID 5013879 Mfr # B-MO-K-8
COMPARE
1/2 Quick-Disconnect Insert x Hose Barb Non-Valved
£5.78GBP / Each
£6.94GBP (incl VAT)
Eldon James SeriesLock® Quick-Disconnect Fitting, Hose Barb Insert, Kynar PVDF, Non-Valved, Large, 5/8" ID 5013880 Mfr # B-MO-K-10
COMPARE
5/8 Quick-Disconnect Insert x Hose Barb Non-Valved
£5.26GBP / Each
£6.31GBP (incl VAT)
Eldon James SeriesLock® Quick-Disconnect Fitting, Hose Barb Insert, Kynar PVDF, Non-Valved, Large, 3/4" ID 5013881 Mfr # B-MO-K-12
COMPARE
3/4 Quick-Disconnect Insert x Hose Barb Non-Valved
£5.41GBP / Each
£6.49GBP (incl VAT)
Eldon James SeriesLock® Quick-Disconnect Fitting, Hose Barb Insert, Panel Mount, Kynar PVDF, Valved, Viton O-Ring, SS Spring, Large, 1/2" ID 5013893 Mfr # PM-MV-K-VS-8
COMPARE
1/2 Quick-Disconnect Panel-Mount Insert x Hose Barb Valved
£18.15GBP / Each
£21.78GBP (incl VAT)
Eldon James SeriesLock® Quick-Disconnect Fitting, Hose Barb Insert, Panel Mount, Kynar PVDF, Valved, Viton O-Ring, SS Heavy Spring, Large, 1/2" ID 5013894 Mfr # PM-MV-K-VS-8H
COMPARE
1/2 Quick-Disconnect Panel-Mount Insert x Hose Barb, Heavy Spring Valved
£24.30GBP / Each
£29.16GBP (incl VAT)
Eldon James SeriesLock® Quick-Disconnect Fitting, Hose Barb Insert, Panel Mount, Kynar PVDF, Valved, Viton O-Ring, SS Spring, Large, 5/8" ID 5013895 Mfr # PM-MV-K-VS-10
COMPARE
5/8 Quick-Disconnect Panel-Mount Insert x Hose Barb Valved
£18.15GBP / Each
£21.78GBP (incl VAT)
Eldon James SeriesLock® Quick-Disconnect Fitting, Hose Barb Insert, Panel Mount, Kynar PVDF, Valved, Viton O-Ring, SS Heavy Spring, Large, 5/8" ID 5013896 Mfr # PM-MV-K-VS-10H
COMPARE
5/8 Quick-Disconnect Panel-Mount Insert x Hose Barb, Heavy Spring Valved
£24.67GBP / Each
£29.60GBP (incl VAT)
Eldon James SeriesLock® Quick-Disconnect Fitting, Hose Barb Insert, Panel Mount, Kynar PVDF, Valved, Viton O-Ring, SS Spring, Large, 3/4" ID 5013897 Mfr # PM-MV-K-VS-12
COMPARE
3/4 Quick-Disconnect Panel-Mount Insert x Hose Barb Valved
£18.52GBP / Each
£22.22GBP (incl VAT)
Eldon James SeriesLock® Quick-Disconnect Fitting, Hose Barb Insert, Panel Mount, Kynar PVDF, Valved, Viton O-Ring, SS Heavy Spring, Large, 3/4" ID 5013898 Mfr # PM-MV-K-VS-12H
COMPARE
3/4 Quick-Disconnect Panel-Mount Insert x Hose Barb, Heavy Spring Valved
£24.67GBP / Each
£29.60GBP (incl VAT)
Eldon James SeriesLock® Quick-Disconnect Fitting, Sanitary Flange Insert, Kynar PVDF, Valved, Viton O-Ring, SS Spring, Large, 3/4" Sanitary & 1/2" ID 5013929 Mfr # SF12-MV-K-VS-8
COMPARE
1/2 Quick-Disconnect Insert x 3/4" Sanitary Flange Valved
£22.38GBP / Each
£26.86GBP (incl VAT)
Eldon James SeriesLock® Quick-Disconnect Fitting, Sanitary Flange Insert, Kynar PVDF, Valved, Viton O-Ring, SS Heavy Spring, Large, 3/4" Sanitary & 1/2" ID 5013930 Mfr # SF12-MV-K-VS-8H
COMPARE
1/2 Quick-Disconnect Insert x 3/4" Sanitary Flange, Heavy Spring Valved
£28.52GBP / Each
£34.22GBP (incl VAT)
Eldon James SeriesLock® Quick-Disconnect Fitting, Sanitary Flange Insert, Kynar PVDF, Valved, Viton O-Ring, SS Spring, Large, 3/4" Sanitary & 5/8" ID 5013931 Mfr # SF12-MV-K-VS-10
COMPARE
5/8 Quick-Disconnect Insert x 3/4" Sanitary Flange Valved
£22.38GBP / Each
£26.86GBP (incl VAT)
Eldon James SeriesLock® Quick-Disconnect Fitting, Sanitary Flange Insert, Kynar PVDF, Valved, Viton O-Ring, SS Heavy Spring, Large, 3/4" Sanitary & 5/8" ID 5013932 Mfr # SF12-MV-K-VS-10H
COMPARE
5/8 Quick-Disconnect Insert x 3/4" Sanitary Flange, Heavy Spring Valved
£28.52GBP / Each
£34.22GBP (incl VAT)
Eldon James SeriesLock® Quick-Disconnect Fitting, Sanitary Flange Insert, Kynar PVDF, Valved, Viton O-Ring, SS Spring, Large, 3/4" Sanitary & 3/4" ID 5013933 Mfr # SF12-MV-K-VS-12
COMPARE
3/4 Quick-Disconnect Insert x 3/4" Sanitary Flange Valved
£22.38GBP / Each
£26.86GBP (incl VAT)
Showing 1 - 20 of 30