Eldon James SeriesLock® Quick-Disconnect Fittings, Large Hose Barb & Sanitary Flange Body, Polysulfone
Specially designed quick-disconnect fitting provides a high-volume flow at low line pressures
  • Spring-free flow path® design
  • Audible click ensures secure connection
  • Safety slide release minimizes accidental disconnects
  • Sterile quick disconnect
  • High flow yet compact to fit in tight spaces
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
The SeriesLock® quick disconnect fittings with spring-free fluid path allow a high-volume flow while minimizing turbulence, foaming, and bubbles. For a secure, leak-free connection, push a large fitting insert into a large fitting body until you hear an audible “click.” Simply use the safety slide release to disconnect the fittings. The slide release eliminates accidental separation while offering easy disconnect with one hand even while wearing gloves. Recessed connection features protect contact surfaces and prolong the life of the O-rings.
Constructed of translucent polysulfone, fittings show excellent resistance to mineral acids, akali and salt solutions, detergents, and hydrocarbon oils. These durable fittings are rated for continuous use at temperatures up to 149°C (300°F).
£12.38 - £33.19GBP / Each
£14.86 - £39.83GBP (incl VAT)
18 variations of this product are available.
See All Product Options Below
18 PRODUCT OPTIONS
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Connection Type Valve
Availability Pricing
Eldon James SeriesLock® Quick-Disconnect Fitting, Hose Barb Body, Polysulfone, Valved, Silicone O-Ring, SS Spring, Large, 1/2" ID 5013834 Mfr # B-FV-SO-SS-8
COMPARE
1/2 Quick-Disconnect Body x Hose Barb Valved
£23.93GBP / Each
£28.72GBP (incl VAT)
Eldon James SeriesLock® Quick-Disconnect Fitting, Hose Barb Body, Polysulfone, Valved, Silicone O-Ring, SS Spring, Large, 5/8" ID 5013835 Mfr # B-FV-SO-SS-10
COMPARE
5/8 Quick-Disconnect Body x Hose Barb Valved
£23.93GBP / Each
£28.72GBP (incl VAT)
Eldon James SeriesLock® Quick-Disconnect Fitting, Hose Barb Body, Polysulfone, Valved, Silicone O-Ring, SS Spring, Large, 3/4" ID 5013836 Mfr # B-FV-SO-SS-12
COMPARE
3/4 Quick-Disconnect Body x Hose Barb Valved
£23.93GBP / Each
£28.72GBP (incl VAT)
Eldon James SeriesLock® Quick-Disconnect Fitting, Hose Barb Body, Polysulfone, Non-Valved, Silicone O-Ring, SS Spring, Large, 1/2" ID 5013849 Mfr # B-FO-SO-SS-8
COMPARE
1/2 Quick-Disconnect Body x Hose Barb Non-Valved
£12.38GBP / Each
£14.86GBP (incl VAT)
Eldon James SeriesLock® Quick-Disconnect Fitting, Hose Barb Body, Polysulfone, Non-Valved, Silicone O-Ring, SS Spring, Large, 5/8" ID 5013850 Mfr # B-FO-SO-SS-10
COMPARE
5/8 Quick-Disconnect Body x Hose Barb Non-Valved
£12.38GBP / Each
£14.86GBP (incl VAT)
Eldon James SeriesLock® Quick-Disconnect Fitting, Hose Barb Body, Polysulfone, Non-Valved, Silicone O-Ring, SS Spring, Large, 3/4" ID 5013851 Mfr # B-FO-SO-SS-12
COMPARE
3/4 Quick-Disconnect Body x Hose Barb Non-Valved
£12.52GBP / Each
£15.02GBP (incl VAT)
Eldon James SeriesLock® Quick-Disconnect Fitting, Sanitary Flange Body, Polysulfone, Valved, Silicone O-Ring, SS Spring, Large, 3/4" Sanitary & 1/2" ID 5013901 Mfr # SF12-FV-SO-SS-8
COMPARE
1/2 Quick-Disconnect Body x 3/4" Sanitary Flange Valved
£31.63GBP / Each
£37.96GBP (incl VAT)
Eldon James SeriesLock® Quick-Disconnect Fitting, Sanitary Flange Body, Polysulfone, Valved, Silicone O-Ring, SS Spring, Large, 3/4" Sanitary & 5/8" ID 5013902 Mfr # SF12-FV-SO-SS-10
COMPARE
5/8 Quick-Disconnect Body x 3/4" Sanitary Flange Valved
£31.63GBP / Each
£37.96GBP (incl VAT)
Eldon James SeriesLock® Quick-Disconnect Fitting, Sanitary Flange Body, Polysulfone, Valved, Silicone O-Ring, SS Spring, Large, 3/4" Sanitary & 3/4" ID 5013903 Mfr # SF12-FV-SO-SS-12
COMPARE
3/4 Quick-Disconnect Body x 3/4" Sanitary Flange Valved
£31.63GBP / Each
£37.96GBP (incl VAT)
Eldon James SeriesLock® Quick-Disconnect Fitting, Sanitary Flange Body, Polysulfone, Non-Valved, Silicone O-Ring, SS Spring, Large, 3/4" Sanitary & 1/2" ID 5013914 Mfr # SF12-FO-SO-SS-8
COMPARE
1/2 Quick-Disconnect Body x 3/4" Sanitary Flange Non-Valved
£24.67GBP / Each
£29.60GBP (incl VAT)
Eldon James SeriesLock® Quick-Disconnect Fitting, Sanitary Flange Body, Polysulfone, Non-Valved, Silicone O-Ring, SS Spring, Large, 3/4" Sanitary & 5/8" ID 5013915 Mfr # SF12-FO-SO-SS-10
COMPARE
5/8 Quick-Disconnect Body x 3/4" Sanitary Flange Non-Valved
£24.67GBP / Each
£29.60GBP (incl VAT)
Eldon James SeriesLock® Quick-Disconnect Fitting, Sanitary Flange Body, Polysulfone, Non-Valved, Silicone O-Ring, SS Spring, Large, 3/4" Sanitary & 3/4" ID 5013916 Mfr # SF12-FO-SO-SS-12
COMPARE
3/4 Quick-Disconnect Body x 3/4" Sanitary Flange Non-Valved
£16.97GBP / Each
£20.36GBP (incl VAT)
Eldon James SeriesLock® Quick-Disconnect Fitting, Sanitary Flange Body, Polysulfone, Valved, Silicone O-Ring, SS Spring, Large, 1-1/2" Sanitary & 1/2" ID 5013945 Mfr # SF24-FV-SO-SS-8
COMPARE
1/2 Quick-Disconnect Body x 1-1/2" Sanitary Flange Valved
£33.19GBP / Each
£39.83GBP (incl VAT)
Eldon James SeriesLock® Quick-Disconnect Fitting, Sanitary Flange Body, Polysulfone, Valved, Silicone O-Ring, SS Spring, Large, 1-1/2" Sanitary & 5/8" ID 5013946 Mfr # SF24-FV-SO-SS-10
COMPARE
5/8 Quick-Disconnect Body x 1-1/2" Sanitary Flange Valved
£33.19GBP / Each
£39.83GBP (incl VAT)
Eldon James SeriesLock® Quick-Disconnect Fitting, Sanitary Flange Body, Polysulfone, Valved, Silicone O-Ring, SS Spring, Large, 1-1/2" Sanitary & 3/4" ID 5013947 Mfr # SF24-FV-SO-SS-12
COMPARE
3/4 Quick-Disconnect Body x 1-1/2" Sanitary Flange Valved
£33.19GBP / Each
£39.83GBP (incl VAT)
Eldon James SeriesLock® Quick-Disconnect Fitting, Sanitary Flange Body, Polysulfone, Non-Valved, Silicone O-Ring, SS Spring, Large, 1-1/2" Sanitary & 1/2" ID 5013954 Mfr # SF24-FO-SO-SS-8
COMPARE
1/2 Quick-Disconnect Body x 1-1/2" Sanitary Flange Non-Valved
£25.49GBP / Each
£30.59GBP (incl VAT)
Eldon James SeriesLock® Quick-Disconnect Fitting, Sanitary Flange Body, Polysulfone, Non-Valved, Silicone O-Ring, SS Spring, Large, 1-1/2" Sanitary & 5/8" ID 5013955 Mfr # SF24-FO-SO-SS-10
COMPARE
5/8 Quick-Disconnect Body x 1-1/2" Sanitary Flange Non-Valved
£25.49GBP / Each
£30.59GBP (incl VAT)
Eldon James SeriesLock® Quick-Disconnect Fitting, Sanitary Flange Body, Polysulfone, Non-Valved, Silicone O-Ring, SS Spring, Large, 1-1/2" Sanitary & 3/4" ID 5013956 Mfr # SF24-FO-SO-SS-12
COMPARE
3/4 Quick-Disconnect Body x 1-1/2" Sanitary Flange Non-Valved
£25.49GBP / Each
£30.59GBP (incl VAT)
Showing 1- 18 of 18