Eldon James Hose Barb Plugs, Natural Nylon
Simply insert the hose barb plug in the soft tubing to stop the fluid flow
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
  • Quick and easy assembly without tools
  • Single barb design tightly “grips” the tubing
  • RoHS compliant
Use these high-quality PVC-free fittings in biomedical research, pharmaceutical, electronic, and other general-purpose applications. A single barb on one end of the fitting creates a tight seal to keep the tubing in place.
These natural nylon fittings provide good tensile strength, stiffness, and impact resistance and are resistant to fuels and lubricants. Not recommended for use with phenols, strong acids, and oxidizing agents.
£5.12 - £17.78GBP / Pkg of 10
£6.14 - £21.34GBP (incl VAT)
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Fitting Material Connection Type
Availability Pricing
Eldon James Hose Barb to Plug Fitting, Straight Adapter, Natural Nylon, 3/32" ID; 10/Pk 5012591 Mfr # P0-1.5-200NN
COMPARE
3/32 Nylon Hose Barb x Plug
£5.12GBP / Pkg of 10
£6.14GBP (incl VAT)
Eldon James Hose Barb to Plug Fitting, Straight Adapter, Natural Nylon, 5/8" ID; 10/Pk 5012592 Mfr # P0-10NN
COMPARE
5/8 Nylon Hose Barb x Plug
£17.78GBP / Pkg of 10
£21.34GBP (incl VAT)
Eldon James Hose Barb to Plug Fitting, Straight Adapter, Natural Nylon, 1/16" ID; 10/Pk 5012593 Mfr # P0-1-200NN
COMPARE
1/16 Nylon Hose Barb x Plug
£5.12GBP / Pkg of 10
£6.14GBP (incl VAT)
Eldon James Hose Barb to Plug Fitting, Straight Adapter, Natural Nylon, 3/4" ID; 10/Pk 5012594 Mfr # P0-12NN
COMPARE
3/4 Nylon Hose Barb x Plug
£12.75GBP / Pkg of 10
£15.30GBP (incl VAT)
Eldon James Hose Barb to Plug Fitting, Straight Adapter, Natural Nylon, 1" ID; 10/Pk 5012595 Mfr # P0-16NN
COMPARE
1 Nylon Hose Barb x Plug
£14.52GBP / Pkg of 10
£17.42GBP (incl VAT)
Eldon James Hose Barb to Plug Fitting, Straight Adapter, Natural Nylon, 5/32" ID; 10/Pk 5012596 Mfr # P0-2.5-200NN
COMPARE
5/32 Nylon Hose Barb x Plug
£6.45GBP / Pkg of 10
£7.74GBP (incl VAT)
Eldon James Hose Barb to Plug Fitting, Straight Adapter, Natural Nylon, 1/8" ID; 10/Pk 5012597 Mfr # P0-2-200NN
COMPARE
1/8 Nylon Hose Barb x Plug
£5.12GBP / Pkg of 10
£6.14GBP (incl VAT)
Eldon James Hose Barb to Plug Fitting, Straight Adapter, Natural Nylon, 3/16" ID; 10/Pk 5012598 Mfr # P0-3-200NN
COMPARE
3/16 Nylon Hose Barb x Plug
£8.52GBP / Pkg of 10
£10.22GBP (incl VAT)
Eldon James Hose Barb to Plug Fitting, Straight Adapter, Natural Nylon, 1/4" ID; 10/Pk 5012599 Mfr # P0-4NN
COMPARE
1/4 Nylon Hose Barb x Plug
£8.97GBP / Pkg of 10
£10.76GBP (incl VAT)
Eldon James Hose Barb to Plug Fitting, Straight Adapter, Natural Nylon, 3/8" ID; 10/Pk 5012600 Mfr # P0-6NN
COMPARE
3/8 Nylon Hose Barb x Plug
£10.67GBP / Pkg of 10
£12.80GBP (incl VAT)
Eldon James Hose Barb to Plug Fitting, Straight Adapter, Natural Nylon, 1/2" ID; 10/Pk 5012601 Mfr # P0-8NN
COMPARE
1/2 Nylon Hose Barb x Plug
£11.49GBP / Pkg of 10
£13.79GBP (incl VAT)
Showing 1- 11 of 11