Eldon James Hose Barb Plugs, Natural Kynar®
Single barb design tightly “grips” tubing
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
  • Biopharma compatible and USP 661
  • BPOG compliant
Use these high-quality PVC-free fittings in biomedical research, pharmaceutical, electronic, and other critical-use applications. Simply insert the hose barb plug in the soft tubing to stop the fluid flow. Each fitting features a single barb on one end, ensuring a tight seal to keep the tubing securely in place.
These natural Kynar® fittings provide excellent chemical, flame, gamma radiation, UV, and weather resistance. They also exhibit good mechanical properties, low moisture absorption, and good machinability.
£8.52 - £19.41GBP / Pkg of 10
£10.22 - £23.29GBP (incl VAT)
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Fitting Material Connection Type
Availability Pricing
Eldon James Hose Barb to Plug Fitting, Straight Adapter, Natural Kynar®, 3/32" ID; 10/PK 5012861 Mfr # P0-1.5-200NK7
COMPARE
3/32 PVDF Hose Barb x Plug
£8.97GBP / Pkg of 10
£10.76GBP (incl VAT)
Eldon James Hose Barb to Plug Fitting, Straight Adapter, Natural Kynar®, 5/8" ID; 10/PK 5012862 Mfr # P0-10NK7
COMPARE
5/8 PVDF Hose Barb x Plug
£17.78GBP / Pkg of 10
£21.34GBP (incl VAT)
Eldon James Hose Barb to Plug Fitting, Straight Adapter, Natural Kynar®, 1/16" ID; 10/PK 5012863 Mfr # P0-1-200NK7
COMPARE
1/16 PVDF Hose Barb x Plug
£8.52GBP / Pkg of 10
£10.22GBP (incl VAT)
Eldon James Hose Barb to Plug Fitting, Straight Adapter, Natural Kynar®, 3/4" ID; 10/PK 5012864 Mfr # P0-12NK7
COMPARE
3/4 PVDF Hose Barb x Plug
£17.78GBP / Pkg of 10
£21.34GBP (incl VAT)
Eldon James Hose Barb to Plug Fitting, Straight Adapter, Natural Kynar®, 1" ID; 10/PK 5012865 Mfr # P0-16NK7
COMPARE
1 PVDF Hose Barb x Plug
£19.41GBP / Pkg of 10
£23.29GBP (incl VAT)
Eldon James Hose Barb to Plug Fitting, Straight Adapter, Natural Kynar®, 5/32" ID; 10/PK 5012866 Mfr # P0-2.5-200NK7
COMPARE
5/32 PVDF Hose Barb x Plug
£10.15GBP / Pkg of 10
£12.18GBP (incl VAT)
Eldon James Hose Barb to Plug Fitting, Straight Adapter, Natural Kynar®, 1/8" ID; 10/PK 5012868 Mfr # P0-2-200NK7
COMPARE
1/8 PVDF Hose Barb x Plug
£8.97GBP / Pkg of 10
£10.76GBP (incl VAT)
Eldon James Hose Barb to Plug Fitting, Straight Adapter, Natural Kynar®, 3/16" ID; 10/PK 5012869 Mfr # P0-3-200NK7
COMPARE
3/16 PVDF Hose Barb x Plug
£11.93GBP / Pkg of 10
£14.32GBP (incl VAT)
Eldon James Hose Barb to Plug Fitting, Straight Adapter, Natural Kynar®, 1/4" ID; 10/PK 5012870 Mfr # P0-4NK7
COMPARE
1/4 PVDF Hose Barb x Plug
£13.93GBP / Pkg of 10
£16.72GBP (incl VAT)
Eldon James Hose Barb to Plug Fitting, Straight Adapter, Natural Kynar®, 3/8" ID; 10/PK 5012871 Mfr # P0-6NK7
COMPARE
3/8 PVDF Hose Barb x Plug
£16.08GBP / Pkg of 10
£19.30GBP (incl VAT)
Eldon James Hose Barb to Plug Fitting, Straight Adapter, Natural Kynar®, 1/2" ID; 10/PK 5012872 Mfr # P0-8NK7
COMPARE
1/2 PVDF Hose Barb x Plug
£16.89GBP / Pkg of 10
£20.27GBP (incl VAT)
Showing 1- 11 of 11