Economical Storage Boxes
Polypropylene (PP) storage boxes are sturdy and won’t deteriorate when exposed to moisture and temperatures down to –80°C (–112°F). These boxes are excellent substitutes for cardboard boxes that tend to degrade and absorb moisture after prolonged use. Translucent plastic lids let you easily identify samples. Boxes accommodate microcentrifuge tubes and cryogenic tubes of various lengths. Autoclavable.
£49.04 - £74.51GBP / Pkg of 6
£58.85 - £89.41GBP (incl VAT)
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubes Held Tube Size
Availability Pricing
COMPARE
50 0.5 mL
£49.04GBP / Pkg of 6
£58.85GBP (incl VAT)
COMPARE
50 1.5 mL
£62.31GBP / Pkg of 6
£74.77GBP (incl VAT)
COMPARE
100 1.5 mL
£74.51GBP / Pkg of 6
£89.41GBP (incl VAT)