Drummond Portable Pipet-Aid XP
The lighter, more compact portable Pipet-Aid
  • Three speed settings provide more precise control for both aspirate and dispense modes
  • EZ Grip™ nosepiece with double membrane filter — prevents use if it becomes wet
  • Includes removable stand and holster-type wall bracket for storage while not in use
These portable Pipet-Aid XP pipette fillers are designed to minimize fatigue. Use with 1 to 100-mL pipettes. Easy two-button operation for independent control of filling or emptying. Unit offers three fill/empty speeds (fast, medium, or slow) that are controlled by the amount each button is depressed. A gravity drain model is available which substitutes a gravity drain function for the fast mode on the dispense button to provide more precise aliquot delivery with volumetric pipets.
£464.60GBP / Dozen of 1
£557.52GBP (incl VAT)
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Material Power (VAC) Description
Availability Pricing
Drummond 4-000-102 Portable Pipet-Aid XP with Gravity Drain 110 V, Sold 1 EA, NOT DZ 0789744 Mfr # 4-000-102
COMPARE
Portable Pipet-Aid XP with Gravity Drain 110 V, sold 1 each, NOT DZ
£464.60GBP / Dozen of 1
£557.52GBP (incl VAT)