Diversey™ Virex® All-Purpose Disinfectant Cleaner
Cleans and deodorizes all in one step
  • Disinfects surfaces in three minutes
  • Kills Norovirus, VRE, and MRSA
  • Ready-to-use one-step disinfectant saves you time
  • Bactericide, tuberculocide, virucide, and fungicide sanitizer
  • Good for cleaning acrylic and polycarbonate barriers
Virex® all-purpose, quaternary-based disinfectant cleaner and deodorizer is ready to use out of the bottle – no diluting needed. Easily apply the cleaner with a mop, sponge, or cloth to hard, inanimate, nonporous surfaces such as floors, walls, porcelain, and plastic surfaces. Ideal for use in hospitals, nursing homes, schools, colleges, and hotels.
Meets OSHA’s bloodborne pathogen standards for decontaminating blood and body fluids. Effective in killing a broad spectrum of microorganisms including HBV, HIV-1, VRE, MRSA, MRSE, VISA, PRSP, Herpes Simplex types 1 & 2, Influenza Type A2, Adenovirus, Rotavirus, and Norovirus.
Also kills organisms that cause odors and works as an odor counteractant with a fresh citrus scent.
£40.32GBP / Case of 8
£48.38GBP (incl VAT)
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Applications Dilution Format
Availability Pricing
Diversey Virex® All Purpose Disinfectant Cleaner, 32 oz Spray Bottles; 8/Case 7890107 Mfr # CBD540533
COMPARE
Bactericide, tuberculocide, virucide, and fungicide sanitizer Ready to use Clear, liquid
£40.32GBP / Case of 8
£48.38GBP (incl VAT)