Dilvac® Blue Enameled Steel Dewar Flasks
Ideal for cold trap
  • Safe for use with all liquid gases
  • Ultra-low evacuated, high-quality borosilicate glass inner flasks
  • Tough blue stove-enameled steel outer container
  • Rust and corrosion resistant
  • Cushioned base prevents benchtop damage
£204.36 - £353.40GBP / Each
£245.23 - £424.08GBP (incl VAT)
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Depth (in) Inside Diameter (ID) (in) Volume (Liters)
Availability Pricing
COMPARE
2 3/4 1 1/2 0.2
£204.36GBP / Each
£245.23GBP (incl VAT)
COMPARE
12 3/8 2 1/8 0.7
£233.43GBP / Each
£280.12GBP (incl VAT)
COMPARE
9 1/2 2 1/2 0.75
£210.87GBP / Each
£253.04GBP (incl VAT)
COMPARE
9 1/4 3 1.1
£224.97GBP / Each
£269.96GBP (incl VAT)
COMPARE
9 1/2 3 1/2 1.5
£275.74GBP / Each
£330.89GBP (incl VAT)
COMPARE
9 1/2 4 1.5
£292.44GBP / Each
£350.93GBP (incl VAT)
COMPARE
11 3/8 4 2
£305.68GBP / Each
£366.82GBP (incl VAT)
COMPARE
9 5 3/8 2
£353.40GBP / Each
£424.08GBP (incl VAT)
Showing 1- 8 of 8