Digi-Sense Thermocouple High-Temperature Wire Probes - 12L, Fiberglass Insulated with Stripped Leads
High-Temperature Thermocouple Probes Designed for Multiple Bends in any Environment
  • Bend probes around pipe diameters as small as twice the sheath diameter to easily determine your pipe temperature.
  • Response Time: 0.85 seconds or greater
  • Type K and J feature ICONEL 600 sheath
  • Type T 316 Stainless Steel sheath
  • Probes are tightly packed with powdered MgO in 12" long sheaths.
  • Probes include 3-ft stranded 24 gauge (0.020" dia) fiberglass wire with stainless steel over braid.
  • Each probe comes with color coded ANSI miniconnectors: type J-black, type K-yellow, type T-blue, and type E-purple
  • Custom designs or changes to this product are available upon request.
Note: the temperature range is for the probe tip only. The handle material may be affected if the probe is rated higher than the handle depending on exposure time, application, and handle material. ABS handles are rated to 220°F (104°C); PVC handles are rated to 200°F (93°C); Stainless Steel handles are rated at 450°F (232°C); Mini Connectors are rated to 392°F (200°C).
£61.52 - £68.61GBP / Each
£73.82 - £82.33GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Probe Type Description
Availability Pricing
COMPARE
1290 K Type-K, Hi-Temperature 12" L, .040" Dia Probe, Sripped Ends, Grounded, 3ft 24-Ga
£61.52GBP / Each
£73.82GBP (incl VAT)
COMPARE
1290 K Type-K, Hi-Temperature 12" L, .040" Dia Probe, Sripped Ends, Ungrd, 3ft 24-Ga
£68.61GBP / Each
£82.33GBP (incl VAT)