Digi-Sense Thermocouple Fine-Gauge Wire Probes
Fine-Gauge Wire Probes feature beaded junctions and 12" leads with very quick response times.
  • Negative lead is slightly shorter than the positive lead for easy identification.
  • Probes come in packs of five.
  • Connectors sold separately.
  • Custom designs or changes to this product are available upon request
Probes with a diameters of 0.005" or 0.010" provide the quickest time constant typically less than 0.3 seconds.
Probes with a diameters of 0.020" or 0.032" has a slower time constant typically 1 second or less but have greater long-term durability.
All probes are uninsulated and are not recommended for use with aqueous solutions (Keep probe wire from touching to prevent a false junction).
£39.12 - £46.37GBP / Pkg of 5
£46.94 - £55.64GBP (incl VAT)
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Probe Type
Availability Pricing
COMPARE
1800 K
£45.88GBP / Pkg of 5
£55.06GBP (incl VAT)
COMPARE
1800 K
£46.37GBP / Pkg of 5
£55.64GBP (incl VAT)
COMPARE
1800 K
£40.91GBP / Pkg of 1
£49.09GBP (incl VAT)
COMPARE
1800 K
£39.12GBP / Pkg of 1
£46.94GBP (incl VAT)
COMPARE
900 J
£40.91GBP / Pkg of 1
£49.09GBP (incl VAT)
COMPARE
900 J
£40.01GBP / Pkg of 5
£48.01GBP (incl VAT)
COMPARE
900 J
£40.91GBP / Pkg of 5
£49.09GBP (incl VAT)
COMPARE
500 T
£40.91GBP / Pkg of 1
£49.09GBP (incl VAT)
COMPARE
500 T
£40.01GBP / Pkg of 1
£48.01GBP (incl VAT)
COMPARE
500 T
£40.91GBP / Pkg of 5
£49.09GBP (incl VAT)
Showing 1- 10 of 10