Digi-Sense Thermocouple Detachable Probe with Locking Handle Kits
Ideal for switching out probes when working with multiple applications.
  • Each probe and handle features a self-locking clip to ensure a sturdy hold onto the handle.
  • Response Time: 1.75 - 3 seconds or greater
  • Each probe comes with a color coded ANSI mini connector: type J-black, type K-yellow, type T-blue, and type E-purple.
  • Custom designs or changes to this product are available upon request
Each kit comes with a quick change handle and three 8” long detachable thermocouples - one general purpose tip, one air/gas tip, and one penetration tip.
Note: the temperature range is for the probe tip only. The handle material may be affected if the probe is rated higher than the handle depending on exposure time, application, and handle material. ABS handles are rated to 220°F (104°C); PVC handles are rated to 200°F (93°C); Stainless Steel handles are rated at 450°F (232°C); Mini Connectors are rated to 392°F (200°C).
£192.50 - £199.63GBP / Each
£231.00 - £239.56GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Probe Type
Availability Pricing
COMPARE
200 K
£199.63GBP / Each
£239.56GBP (incl VAT)
COMPARE
200 T
£192.50GBP / Each
£231.00GBP (incl VAT)