Digi-Sense Temperature Probe Compression Fittings
Fittings let you adjust the depth your probe extends into a media you are measuring
  • Available in both 316 Stainless Steel and Brass
Use fittings for straight-shaft probes and metal tubing only. Tighten compression nut to secure and completely seal probe. Replacement ferrules let you reuse a compression fitting when replacing a worn-out probe. Simply disassemble fitting, discard probe and old ferrule, install replacement ferrule over probe sheath, and retighten fitting. Connectors come in both 316 Stainless Steel and Brass.
£11.66 - £24.12GBP / Each
£13.99 - £28.94GBP (incl VAT)
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Compression Fitting; Probe Diameter 1/8"; 316 Stainless Steel; 1/8"NPT (M)
£18.01GBP / Each
£21.61GBP (incl VAT)
COMPARE
Compression Fitting; Probe Diameter 3/16"; 316 Stainless Steel; 1/8"NPT (M)
£18.01GBP / Each
£21.61GBP (incl VAT)
COMPARE
Compression Fitting; Probe Diameter 3/16"; 316 Stainless Steel; 1/4"NPT (M)
£18.01GBP / Each
£21.61GBP (incl VAT)
COMPARE
Compression Fitting; Probe Diameter 1/4"; 316 Stainless Steel 1/8"NPT (M)
£17.03GBP / Each
£20.44GBP (incl VAT)
COMPARE
Compression Fitting; Probe Diameter 1/4"; 316 Stainless Steel 1/4"NPT (M)
£21.11GBP / Each
£25.33GBP (incl VAT)
COMPARE
Compression Fitting; Probe Diameter 3/8"; 316 Stainless Steel 3/8"NPT (M)
£24.12GBP / Each
£28.94GBP (incl VAT)
COMPARE
Compression Fitting; Probe Diameter 1/4"; Brass; 1/8"NPT (M)
£11.66GBP / Each
£13.99GBP (incl VAT)
COMPARE
Compression Fitting; Probe Diameter 1/4"; Brass; 1/4"NPT (M)
£11.66GBP / Each
£13.99GBP (incl VAT)
Showing 1- 8 of 8