Digi-Sense Locking Thermocouple Miniconnectors
No more unwanted disconnections
  • Self-locking plugs and jacks secure connection against vibrations and tugging
  • Glass-filled nylon construction provides rigidity, strength, and surface hardness
These plugs and jacks feature a locking mechanism designed to prevent inadvertent uncoupling. Each flat-blade style connector is ANSI color-coded by thermocouple type: black (J), yellow (K), or blue (T).
£10.26 - £11.12GBP / Each
£12.31 - £13.34GBP (incl VAT)
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Locking Miniconnector, Type-K Thermocouple, Male, 1/Ea
£10.26GBP / Each
£12.31GBP (incl VAT)
COMPARE
Locking Miniconnector, Type-J Thermocouple, Male, 1/Ea
£10.26GBP / Each
£12.31GBP (incl VAT)
COMPARE
Locking Miniconnector, Type-T Thermocouple, Male, 1/Ea
£10.26GBP / Each
£12.31GBP (incl VAT)
COMPARE
Locking Miniconnector, Type-K Thermocouple, Female, 1/Ea
£11.12GBP / Each
£13.34GBP (incl VAT)
COMPARE
Locking Miniconnector, Type-J Thermocouple, Female, 1/Ea
£10.26GBP / Each
£12.31GBP (incl VAT)