Digi-Sense Food Infrared Stick Thermometer
Ideal for food inspections and HACCP programs
  • Unique ergonomic stick-style design fits comfortably in your hand
  • Water- and dust-resistant IP54 rating
  • Laser sighting to pinpoint the center of the measurement area
  • Safely and easily locate hot and cold spots, preventing problems and saving costs
  • Note: As of 6/23/2014 this product was rebranded Digi-Sense
Taking temperature measurements has never been easier-just point the thermometer at your target and push one button! The Digi-Sense infrared Stick thermometer is ideal for use in the food market, but can also be used in the home, office, field, or lab-anywhere you need to quickly and accurately measure surface temperatures.
The Digi-Sense infrared Stick thermometer features a IP-54 water- and dust-resistant rating distance-to-sight ratio of 4:1, temperature range of 0 to 575°F (–18 to 302°C) and ±2% basic accuracy. Other features include automatic data Hold, °F/°C selectable units, fixed emissivity, backlit display, and auto power-off. Accuracy of ±2degF (±1degC) over the HACCP critical temperature range of 40 to 140degF (4 to 60 degC).
£117.75 - £121.62GBP / Each
£141.30 - £145.94GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Distance To Target Size Ratio (D:S) Max Temperature (° F) Description
Availability Pricing
COMPARE
4:1 575 4: 1 Food Infrared Stick Thermometer
£117.75GBP / Each
£141.30GBP (incl VAT)
COMPARE
4:1 575 4: 1 Food Infrared Stick Thermometer with NIST Traceable Calibration
£121.62GBP / Each
£145.94GBP (incl VAT)