CPC® (Colder) PLCD Quick-Disconnect Fittings, Hose Barb Body & Insert, Valved, Acetal & Polypropylene
Resistant to most chemical solutions
  • Ideal for general purposes, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support is available to determine which connector type is needed for your application
  • Ergonomic design makes it easy to connect and disconnect fittings quickly
  • Ideal for tubing sizes from 1/4” to 3/8” ID
The CPC (Colder) PLCD Series quick disconnect hose barb body and insert have a 1/4" flow size, making them suitable for a wide range of general-purpose and chemical applications. The fittings feature an automatic shutoff spring-operated stainless-steel valve which closes and seals the fluid pathway upon decoupling, protecting the sterility on each side of the system.
Constructed from durable acetal thermoplastic, these fittings offer high strength, rigidity across a broad temperature range, and resistance to many mild chemical solutions. Alternatively, polypropylene versions provide enhanced chemical resistance, making them ideal for harsher environments.
An ergonomic thumb latch enables effortless one-handed connection and disconnection, even when wearing gloves. The fittings can withstand pressures from vacuum to 120 psi (8.3 bar).
£6.62 - £346.30GBP / Each
£7.94 - £415.56GBP (incl VAT)
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Fitting Material Flow Pattern
Availability Pricing
CPC (Colder) PLCD12 Quick-Disconnect Fittings, Hose Barb Insert, Polypropylene, Valved, 1/4" ID;  25/Pk 4901473 Mfr # PLCD2200412/25 PK
COMPARE
1/4 Polypropylene Straight
£238.52GBP / Pkg of 25
£286.22GBP (incl VAT)
CPC (Colder) PLCD12 Quick-Disconnect Fittings, Hose Barb Body, Polypropylene, Valved, 3/8" ID; 25Pk 4901537 Mfr # PLCD1700612/25 PK
COMPARE
3/8 Polypropylene Straight
£346.30GBP / Pkg of 25
£415.56GBP (incl VAT)
CPC (Colder) PLCD Quick-Disconnect Fittings, Hose Barb Body, Panel Mount, Acetal, Valved, 1/4" ID 5013612 Mfr # PLCD16004
COMPARE
1/4 Acetal Straight
£16.96GBP / Each
£20.35GBP (incl VAT)
CPC (Colder) PLCD Quick-Disconnect Fittings, Hose Barb Body, Acetal, Valved, 1/4" ID 5013613 Mfr # PLCD17004
COMPARE
1/4 Acetal Straight
£8.72GBP / Each
£10.46GBP (incl VAT)
CPC (Colder) PLCD12 Quick-Disconnect Fittings, Hose Barb Body, Polypropylene, Valved, 1/4" ID 5013614 Mfr # PLCD1700412
COMPARE
1/4 Polypropylene Straight
£15.09GBP / Each
£18.11GBP (incl VAT)
CPC (Colder) PLCD Quick-Disconnect Fittings, Hose Barb Body, Acetal, Valved, 3/8" ID 5013615 Mfr # PLCD17006
COMPARE
3/8 Acetal Straight
£8.64GBP / Each
£10.37GBP (incl VAT)
CPC (Colder) PLCD12 Quick-Disconnect Fittings, Hose Barb Body, Polypropylene, Valved, 3/8" ID 5013616 Mfr # PLCD1700612
COMPARE
3/8 Polypropylene Straight
£16.28GBP / Each
£19.54GBP (incl VAT)
CPC (Colder) PLCD Quick-Disconnect Fittings, Hose Barb Insert, Acetal, Valved, 1/4" ID 5013620 Mfr # PLCD22004
COMPARE
1/4 Acetal Straight
£6.62GBP / Each
£7.94GBP (incl VAT)
CPC (Colder) PLCD12 Quick-Disconnect Fittings, Hose Barb Insert, Polypropylene, Valved, 1/4" ID 5013621 Mfr # PLCD2200412
COMPARE
1/4 Polypropylene Straight
£10.82GBP / Each
£12.98GBP (incl VAT)
CPC (Colder) PLCD Quick-Disconnect Fittings, Hose Barb Insert, Acetal, Valved, 3/8" ID 5013622 Mfr # PLCD22006
COMPARE
3/8 Acetal Straight
£6.83GBP / Each
£8.20GBP (incl VAT)
CPC (Colder) PLCD12 Quick-Disconnect Fittings, Hose Barb Insert, Polypropylene, Valved, 3/8" ID 5013623 Mfr # PLCD2200612
COMPARE
3/8 Polypropylene Straight
£12.87GBP / Each
£15.44GBP (incl VAT)
CPC (Colder) PLCD Quick-Disconnect Fittings, Hose Barb Insert, Elbow, Acetal, Valved, 1/4" ID 5013624 Mfr # PLCD23004
COMPARE
1/4 Acetal Elbow
£11.44GBP / Each
£13.73GBP (incl VAT)
CPC (Colder) PLCD12 Quick-Disconnect Fittings, Hose Barb Insert, Elbow, Polypropylene, Valved, 1/4" ID 5013625 Mfr # PLCD2300412
COMPARE
1/4 Polypropylene Elbow
£14.08GBP / Each
£16.90GBP (incl VAT)
CPC (Colder) PLCD Quick-Disconnect Fittings, Hose Barb Insert, Elbow, Acetal, Valved, 3/8" ID 5013626 Mfr # PLCD23006
COMPARE
3/8 Acetal Elbow
£11.82GBP / Each
£14.18GBP (incl VAT)
Showing 1- 14 of 14