CPC® (Colder) MicroCNX® Series Quick-Disconnect Fittings, Genderless Luer
Micro-flow genderless sterile connectors simplify process integration while maximizing flexibility
  • Simple “pinch, click, and pull” design minimizes operator errors
  • Simplified tubing connection provides high performance and reliability
  • Designed for micro-flow media transfer applications
  • Free technical application support available to determine which connector type is needed for your application
The MicroCNX® genderless luer connectors provide fast, easy, secure connections and quick assembly. The interior of the flow path is never exposed to the exterior environment providing a sterile, closed connection on an existing luer connection. Connectors eliminate the need for tube welding, saving you time and money. To use, simply pinch to remove the protective cover, join two halves together and squeeze until you hear an audible “click”, then pull the membrane strips away from the connector.
These aseptic quick connectors are made from animal-derived component-free materials, making them ideal for a wide range of biopharmaceutical, bioprocessing, cell therapy, gene therapy, and other critical-use applications.
£31.04GBP / Each
£37.25GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Fitting Material Luer Connection Type (Port 1) Max Operating Pressure (PSI)
Availability Pricing
CPC (Colder) MicroCNX® Quick-Disconnect Fitting, Micro-Flow, Genderless Female Luer, Polycarbonate 5013825 Mfr # CNX170LFHT
COMPARE
Polycarbonate Female 43.5
£31.04GBP / Each
£37.25GBP (incl VAT)
CPC (Colder) MicroCNX® Quick-Disconnect Fitting, Micro-Flow, Genderless Male Luer, Polycarbonate 5013826 Mfr # CNX170LMHT
COMPARE
Polycarbonate Male 43.5
£31.04GBP / Each
£37.25GBP (incl VAT)