CPC® (Colder) MicroCNX® Series Quick-Disconnect Fittings, Genderless Hose Barb
Micro-flow genderless sterile connectors simplify process integration while maximizing flexibility
  • Simple “pinch, click, and pull” design minimizes operator errors
  • Simplified tubing connection provides high performance and reliability
  • Designed for micro-flow media transfer applications
  • Free technical application support available to determine which connector type is needed for your application
The MicroCNX® genderless connectors provide fast, easy, secure connections and quick assembly. The interior of the flow path is never exposed to the exterior environment providing sterile connections, even in non-sterile environments. Connectors eliminate the need for tube welding, saving you time and money. To use, simply pinch to remove the protective cover, join two halves together and squeeze until you hear an audible “click”, then pull the membrane strips away from the connector.
These aseptic quick connectors are made from animal-derived component-free materials, making them ideal for a wide range of biopharmaceutical, bioprocessing, cell therapy, gene therapy, and other critical-use applications.
£29.40 - £740.75GBP / Pkg of 25
£35.28 - £888.90GBP (incl VAT)
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Fitting Material Max Operating Pressure (PSI)
Availability Pricing
CPC (Colder) MicroCNX® ULT Quick-Disconnect Fitting, Micro-Flow, Genderless Hose Barb, Polyphenylsulfone, 1/16" ID; 25/PK 4901511 Mfr # CNX17101HT/25 PK
COMPARE
1/16 Polyphenylsulfone 43.5
£740.75GBP / Pkg of 25
£888.90GBP (incl VAT)
CPC (Colder) MicroCNX® ULT Quick-Disconnect Fitting, Micro-Flow, Genderless Hose Barb, Polyphenylsulfone, 1/8" ID; 25/PK 4901512 Mfr # CNX17102HT/25 PK
COMPARE
1/8 Polyphenylsulfone 43.5
£740.75GBP / Pkg of 25
£888.90GBP (incl VAT)
CPC (Colder) MicroCNX® Quick-Disconnect Fitting, Micro-Flow, Genderless Hose Barb, Polycarbonate, 1/16" ID 5011214 Mfr # CNX17001HT
COMPARE
1/16 Polycarbonate 60
£30.98GBP / Each
£37.18GBP (incl VAT)
CPC (Colder) MicroCNX® Quick-Disconnect Fitting, Micro-Flow, Genderless Hose Barb, Polycarbonate, 1/8" ID 5011215 Mfr # CNX17002HT
COMPARE
1/8 Polycarbonate 60
£29.48GBP / Each
£35.38GBP (incl VAT)
CPC (Colder) MicroCNX® Quick-Disconnect Fitting, Micro-Flow, Genderless Hose Barb, Polycarbonate, 3/32" ID 5011216 Mfr # CNX17003HT
COMPARE
3/32 Polycarbonate 60
£29.40GBP / Each
£35.28GBP (incl VAT)
CPC (Colder) MicroCNX® ULT Quick-Disconnect Fitting, Micro-Flow, Genderless Hose Barb, Polyphenylsulfone, 1/16" ID 5013827 Mfr # CNX17101HT
COMPARE
1/16 Polyphenylsulfone 43.5
£34.30GBP / Each
£41.16GBP (incl VAT)
CPC (Colder) MicroCNX® ULT Quick-Disconnect Fitting, Micro-Flow, Genderless Hose Barb, Polyphenylsulfone, 1/8" ID 5013828 Mfr # CNX17102HT
COMPARE
1/8 Polyphenylsulfone 43.5
£34.23GBP / Each
£41.08GBP (incl VAT)
CPC (Colder) MicroCNX® ULT Quick-Disconnect Fitting, Micro-Flow, Genderless Hose Barb, Polyphenylsulfone, 3/32" ID 5013829 Mfr # CNX17103HT
COMPARE
3/32 Polyphenylsulfone 43.5
£34.23GBP / Each
£41.08GBP (incl VAT)
Showing 1- 8 of 8