CPC® (Colder) HFC Quick-Disconnect Fittings, Hose Barb Body, Polysulfone with Alloy C-276 Valve
Valved connectors ensure sterility is maintained on both sides of the system during the disconnection process
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • Connect to a male coupling insert for easy fluid transfer
  • Audible “click” provides added assurance of a secure connection
  • Quick one-step disconnection process minimize user error
  • Alloy C-276 valve spring stops the flow and offers broad application compatibility
  • Laser-etched item and lot numbers offer batch traceability
The CPC (Colder) HFC series connectors feature an automatic shutoff valve enabling quick disconnection while maintaining its sterility. Connect a male insert and a female body for a secure fluid flow. Simply push the thumb latch to disconnect. An alloy C-276 valve spring in each connector closes and seals the fluid pathway upon decoupling, protecting the sterility on each side of the system. Laser-etched item and lot numbers provide batch traceability.
All connectors are made from animal-derived component-free materials, making them ideal for a wide range of biopharmaceutical, bioprocessing, and other critical-use applications.
£40.84 - £40.99GBP / Each
£49.01 - £49.19GBP (incl VAT)
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Connection Type Description
Availability Pricing
CPC (Colder) HFC Quick-Disconnect Fitting, Hose Barb Body, Polysulfone with Alloy C-276 Valve, 1/4" ID; 1/Each 5011223 Mfr # HFCD17439MHC
COMPARE
1/4 Quick Disconnect Body x Hose Barb Quick-Disconnect Fitting, Hose Barb Body, Polysulfone with Alloy C-276 Valve, 1/4" ID; 1/Ea
£40.99GBP / Each
£49.19GBP (incl VAT)
CPC (Colder) HFC Quick-Disconnect Fitting, Hose Barb Body, Polysulfone with Alloy C-276 Valve, 3/8" ID; 1/Each 5011225 Mfr # HFCD17639MHC
COMPARE
3/8 Quick Disconnect Body x Hose Barb Quick-Disconnect Fitting, Hose Barb Body, Polysulfone with Alloy C-276 Valve, 3/8" ID; 1/Ea
£40.84GBP / Each
£49.01GBP (incl VAT)
CPC (Colder) HFC Quick-Disconnect Fitting, Hose Barb Body, Polysulfone with Alloy C-276 Valve, 1/2" ID; 1/Each 5011227 Mfr # HFCD17839MHC
COMPARE
1/2 Quick Disconnect Body x Hose Barb Quick-Disconnect Fitting, Hose Barb Body, Polysulfone with Alloy C-276 Valve, 1/2" ID; 1/Ea
£40.99GBP / Each
£49.19GBP (incl VAT)