Cole-Parmer® Wide-Mouth Plastic Bottles, LDPE
Get better sample visibility
  • Leakproof polypropylene screw closures
  • Withstand temperatures to -148°F (-100°C)
Easy to fill, translucent, flexible, and impact resistant LDPE bottles offer better content visibility than high-density polyethylene (HDPE) and low trace metal content. All have leakproof linerless polypropylene (PP) screw closures, suitable for shipping liquids. Can be used for long-term storage in freezers to –148°F (–100°C).
£34.35 - £81.39GBP / Pkg of 6
£41.22 - £97.67GBP (incl VAT)
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Cap / Stopper Size Material
Availability Pricing
COMPARE
30 28 Low Density Polyethyene (LDPE)
£34.35GBP / Pkg of 12
£41.22GBP (incl VAT)
COMPARE
60 36 Low Density Polyethyene (LDPE)
£35.18GBP / Pkg of 12
£42.22GBP (incl VAT)
COMPARE
125 43 Low Density Polyethyene (LDPE)
£42.23GBP / Pkg of 12
£50.68GBP (incl VAT)
COMPARE
250 43 Low Density Polyethyene (LDPE)
£60.07GBP / Pkg of 12
£72.08GBP (incl VAT)
COMPARE
500 53 Low Density Polyethyene (LDPE)
£81.39GBP / Pkg of 12
£97.67GBP (incl VAT)
COMPARE
1000 63 Low Density Polyethyene (LDPE)
£76.50GBP / Pkg of 6
£91.80GBP (incl VAT)