Cole-Parmer® White-Frosted Microscope Slides, Soda Lime Glass
Suitable for marking with inkjet and thermal transfer printers and permanent markers
  • Frosted one end on one side
  • Chemically resistant white writing surface
  • Coating maintains original color in routine H&E staining
  • Each slide measures 76 x 26 mm and 1.0 to 1.2-mm thick
Slightly rough surface of the slides provides better adhesion for delicate samples as well as makes them easier to handle.
£4.90 - £5.56GBP / Pkg of 50
£5.88 - £6.67GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Slide Type Description
Availability Pricing
COMPARE
White frost, Ground Edges, 45° Corners Microscope Slides, White-Frosted, Ground Edges, 45° Corners; 50/PK
£5.56GBP / Pkg of 50
£6.67GBP (incl VAT)
COMPARE
White frost, Cut Edge, 90° Corners Microscope Slides, White-Frosted, Ground Edges, 90° Corners; 50/PK
£4.90GBP / Pkg of 50
£5.88GBP (incl VAT)