Cole-Parmer® Vortamix Mini Vortex Shaker
Compact and powerful enough for 50-mL centrifuge tubes
  • Press your tube to the top gasket and mix as long as needed
  • Agitate tubes at 3000 rpm with a 4.5 mm orbit—even 50-mL, conical-bottom, centrifuge tubes
  • Choose the gasket color you prefer—red is included; blue, green, and purple are available
Compact vortex shaker mixes samples in centrifuge and test tubes, up to 30 mm in diameter, with little effort. The vortex motion is activated by placing a tube on the gasket cap and pressing down lightly.
£163.38 - £170.46GBP / Each
£196.06 - £204.55GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Mini Vortexer, 100-240 VAC; US Adapter
£163.38GBP / Each
£196.06GBP (incl VAT)
COMPARE
Mini Vortexer, 100-240 VAC; International
£170.46GBP / Each
£204.55GBP (incl VAT)