Cole-Parmer® Venturi Vacuum Pumps with Air Saver Technology
Minimizes compressed air usage –which saves you money!
  • Vacuum level switch shuts off air supply when desired vacuum setting is reached
  • Powered only by compressed air—no batteries or electrical connections
  • Holds vacuum even if air supply is lost
The air saver circuitry is perfect for any lab or process application. Powered by compressed air only; the pneumatic vacuum switch is the brain behind the system that constantly monitors and controls the vacuum level. Eliminates the constant air loss and noise of most venturi vacuum pumps. Since it uses no electricity it is inherently intrinsically safe. The vacuum switch is adjustable from 0 to 30” Hg with a hysteresis of 3”Hg.
£1,348.15 - £1,451.86GBP / Each
£1,617.78 - £1,742.23GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Free-Air Capacity (CFM) Max Vacuum (in Hg)
Availability Pricing
COMPARE
1.2 28
£1,348.15GBP / Each
£1,617.78GBP (incl VAT)
COMPARE
0.2 28
£1,451.86GBP / Each
£1,742.23GBP (incl VAT)