Cole-Parmer® Variable Area Flowmeters, 65 mm, Aluminum Mounting Fitting, Cartridge Valve
Ideal for precise low flow rate measurements
  • Corrosion-resistant aluminum fittings ensure long-term reliability
  • Easy installation and exchange of flow tubes
  • Non-rotating adapter prevents tubes from turning
  • Panel mounting with hex nuts makes installation simple and secure
  • Chemical compatibility
  • 1 year warranty
These single flow tube variable area flowmeters with cartridge valves are designed for precise low flow rate measurements of air and water in mL/minute at a given set of pressure and temperature parameters. The Optigrad™ 65 mm universal millimeter scale minimizes parallax and eye fatigue. Particularly suitable for metering carrier gases in chromatography, indicating and controlling gases in manufacturing processes, liquid and gas measurement in laboratories, and pilot plants.
Built-in high-resolution control valves offer high sensitivity control and resolution in conjunction with metering tubes of very low flow rates. Other features include standard mounting fittings, side plates, thick protective magnifying front shield and back plate, and flow tubes.
£152.23 - £168.96GBP / Each
£182.68 - £202.75GBP (incl VAT)
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
Compareitem specifications
Max Air Flow Rate (mLmin) Max Pressure (psig) Description
Availability Pricing
COMPARE
19 200 Variable Area Flowmeter, 65 mm, Aluminum Fitting, 1/8" NPT(F), 19 mL/min. (Air), Cartridge Valve
£152.23GBP / Each
£182.68GBP (incl VAT)
COMPARE
49 200 Variable Area Flowmeter, 65 mm, Aluminum Fitting, 1/8" NPT(F), 49 mL/min. (Air), 0.5 mL/min. (H2O), Cartridge Valve
£168.96GBP / Each
£202.75GBP (incl VAT)
COMPARE
145 200 Variable Area Flowmeter, 65 mm, Aluminum Fitting, 1/8" NPT(F), 145 mL/min. (Air), 2.3 mL/min. (H2O), Cartridge Valve
£152.23GBP / Each
£182.68GBP (incl VAT)
COMPARE
216 200 Variable Area Flowmeter, 65 mm, Aluminum Fitting, 1/8" NPT(F), 216 mL/min. (Air), 3.0 mL/min. (H2O), Cartridge Valve
£152.23GBP / Each
£182.68GBP (incl VAT)
COMPARE
538 200 Variable Area Flowmeter, 65 mm, Aluminum Fitting, 1/8" NPT(F), 538 mL/min. (Air), 11.7 mL/min. (H2O), Cartridge Valve
£152.23GBP / Each
£182.68GBP (incl VAT)
COMPARE
1036 200 Variable Area Flowmeter, 65 mm, Aluminum Fitting, 1/8" NPT(F), 1,036 mL/min. (Air), 20 mL/min. (H2O), Cartridge Valve
£168.96GBP / Each
£202.75GBP (incl VAT)
COMPARE
2520 200 Variable Area Flowmeter, 65 mm, Aluminum Fitting, 1/8" NPT(F), 2,520 mL/min. (Air), 70.5 mL/min. (H2O), Cartridge Valve
£152.23GBP / Each
£182.68GBP (incl VAT)
COMPARE
4917 200 Variable Area Flowmeter, 65 mm, Aluminum Fitting, 1/8" NPT(F), 4,917 mL/min. (Air), 143 mL/min. (H2O), Cartridge Valve
£152.23GBP / Each
£182.68GBP (incl VAT)
COMPARE
12058 200 Variable Area Flowmeter, 65 mm, Aluminum Fitting, 1/8" NPT(F), 12,058 mL/min. (Air), 364 mL/min. (H2O), Cartridge Valve
£152.23GBP / Each
£182.68GBP (incl VAT)
COMPARE
24452 200 Variable Area Flowmeter, 65 mm, Aluminum Fitting, 1/8" NPT(F), 24,452 mL/min. (Air), 723 mL/min. (H2O), Cartridge Valve
£152.23GBP / Each
£182.68GBP (incl VAT)
Showing 1- 10 of 10