Cole-Parmer® Variable Area Air Flowmeters, Stainless Steel Fittings, No Valves
Our variable area acrylic flowmeters offer precision and durability for a wide range of applications
  • Crystal-clear acrylic material with a stable float ensures easy viewing and long-lasting performance
  • Superior, corrosion-resistant stainless-steel fittings ensure long-term reliability in harsh environments
  • Easily convertible from panel to partial or full in-line mounting configurations to suit your needs
  • Easy-to-read English or metric scales for clear and precise measurement
  • Panel mounting with hex nuts makes installation simple and secure
  • Easy disassembly and reassembly for quick cleaning and maintenance
Whether you need an accurate air flow measurement solution for industrial processes, lab setups, or HVAC systems, our acrylic flowmeters provide the perfect combination of precision, durability, and convenience. With a variety of customizable options, they are designed to fit seamlessly into your workflow. Flowmeters without valves are best suited for applications where the flow rate remains constant or is controlled by another device.
£81.49 - £90.44GBP / Each
£97.79 - £108.53GBP (incl VAT)
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Float Max Flow Rate (LPM) Max Flow Rate (SCFH)
Availability Pricing
COMPARE
Glass 1.4 2.8
£81.49GBP / Each
£97.79GBP (incl VAT)
COMPARE
Stainless 2.75 5.5
£81.49GBP / Each
£97.79GBP (incl VAT)
COMPARE
Carboloy 3.5 7
£81.49GBP / Each
£97.79GBP (incl VAT)
COMPARE
Glass 8.5 18
£90.44GBP / Each
£108.53GBP (incl VAT)
COMPARE
Stainless 16 32.5
£81.49GBP / Each
£97.79GBP (incl VAT)
COMPARE
Carboloy 22 45
£81.49GBP / Each
£97.79GBP (incl VAT)
COMPARE
Stainless 50 100
£81.49GBP / Each
£97.79GBP (incl VAT)
Showing 1- 7 of 7