Cole-Parmer® Upright Freezer Sliding Tray Eco-Racks for Standard 2" Boxes
Convenient handles make it easy to pull out the tray while wearing protective gloves
  • Segmented drawers prevent boxes from freezing or sticking together
  • Use with standard 2" cardboard, plastic, or metal boxes
  • Rack frame and sliding tray are constructed of corrosion-resistant stainless steel
These top loading sliding tray eco-racks are a great economical alternative to the standing side access drawer racks. Trays slide out for easy retrieval of samples without having to remove the entire rack from the freezer.
£118.36 - £345.89GBP / Each
£142.03 - £415.07GBP (incl VAT)
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity Height (in) Length (in)
Availability Pricing
COMPARE
3 x 4 Array 8 13/16 16 1/2
£148.71GBP / Each
£178.45GBP (incl VAT)
COMPARE
3 x 5 Array 11 16 1/2
£186.39GBP / Each
£223.67GBP (incl VAT)
COMPARE
4 x 2 Array 4 9/16 22
£118.36GBP / Each
£142.03GBP (incl VAT)
COMPARE
4 x 3 Array 6 5/8 22
£148.71GBP / Each
£178.45GBP (incl VAT)
COMPARE
4 x 4 Array 8 13/16 22
£200.65GBP / Each
£240.78GBP (incl VAT)
COMPARE
4 x 5 Array 11 22
£246.89GBP / Each
£296.27GBP (incl VAT)
COMPARE
5 x 2 Array 4 9/16 26 3/4
Call For Price
COMPARE
5 x 3 Array 6 5/8 26 3/4
£205.75GBP / Each
£246.90GBP (incl VAT)
COMPARE
5 x 4 Array 8 13/16 26 3/4
£273.78GBP / Each
£328.54GBP (incl VAT)
COMPARE
5 x 5 Array 11 26 3/4
£345.89GBP / Each
£415.07GBP (incl VAT)
Showing 1- 10 of 10