Cole-Parmer® Upright Freezer Racks for 100-Place Slide Boxes
Great for storing large quantities of slide boxes
  • Get organized for easier retrieval of samples
  • Accommodates slide boxes measuring up to 8 1/4"L x 7"W x 1 7/16"H
  • Easily access boxes from either side of rack
  • Sturdy corrosion-resistant stainless steel construction for long-term use
Racks feature handles at both ends to accommodate freezers with right- or left-handed doors.
£202.28 - £354.45GBP / Each
£242.74 - £425.34GBP (incl VAT)
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity Height (in) Length (in)
Availability Pricing
COMPARE
3 x 4 Array 6 1/4 21 9/16
£234.26GBP / Each
£281.11GBP (incl VAT)
COMPARE
4 x 4 Array 6 1/4 28 11/16
£286.41GBP / Each
£343.69GBP (incl VAT)
COMPARE
2 x 5 Array 7 13/16 14 3/8
£202.28GBP / Each
£242.74GBP (incl VAT)
COMPARE
3 x 5 Array 7 13/16 21 9/16
£265.23GBP / Each
£318.28GBP (incl VAT)
COMPARE
3 x 6 Array 9 5/16 21 9/16
£290.97GBP / Each
£349.16GBP (incl VAT)
COMPARE
2 x 7 Array 10 7/8 14 3/8
£246.89GBP / Each
£296.27GBP (incl VAT)
COMPARE
3 x 7 Array 10 7/8 21 9/16
£316.56GBP / Each
£379.87GBP (incl VAT)
COMPARE
4 x 7 Array 10 7/8 28 11/16
Call For Price
COMPARE
2 x 8 Array 12 3/8 14 3/8
£273.78GBP / Each
£328.54GBP (incl VAT)
COMPARE
3 x 8 Array 12 3/8 21 9/16
£354.45GBP / Each
£425.34GBP (incl VAT)
Showing 1- 10 of 10