Cole-Parmer® Upright Freezer Drawer Racks for 96/384-Well Microtiter Plates & 96-Well Microtube Boxes
Drawer racks allow easy access to microtiter plates and microtube boxes
  • Quickly store and retrieve plates and boxes without taking out the whole rack
  • Accommodates microtube boxes with 2-1/8" max height
  • Durable corrosion-resistant stainless steel construction for long-term use
Drawer racks facilitate access to samples without having to remove the entire rack from the freezer. Easily retrieve sample boxes by simply sliding out the individual rack drawer.
£139.34 - £402.12GBP / Each
£167.21 - £482.54GBP (incl VAT)
13 variations of this product are available.
See All Product Options Below
13 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity Height (in) Length (in)
Availability Pricing
COMPARE
(27 w/lids or 36 w/o lids) 96/384-well microtiter plates or (9) boxes w/2 1/8" max height 7 5/16 10 15/16
£139.34GBP / Each
£167.21GBP (incl VAT)
COMPARE
(36 w/lids or 48 w/o lids) 96/384-well microtiter plates or (12) boxes w/2 1/8" max height 9 11/16 10 15/16
£178.04GBP / Each
£213.65GBP (incl VAT)
COMPARE
(36 w/lids or 48 w/o lids) 96/384-well microtiter plates or (12) boxes w/2 1/8" max height 7 5/16 14 1/2
£170.50GBP / Each
£204.60GBP (incl VAT)
COMPARE
(48 w/lids or 64 w/o lids) 96/384-well microtiter plates or (16) boxes w/2 1/8" max height 9 11/16 14 1/2
£216.54GBP / Each
£259.85GBP (incl VAT)
COMPARE
(45 w/lids or 60 w/o lids) 96/384-well microtiter plates or (15) boxes w/2 1/8" max height 7 5/16 18
£185.58GBP / Each
£222.70GBP (incl VAT)
COMPARE
(60 w/lids or 80 w/o lids) 96/384-well microtiter plates or (20) boxes w/2 1/8" max height 9 11/16 18
£256.26GBP / Each
£307.51GBP (incl VAT)
COMPARE
(75 w/lids or 100 w/o lids) 96/384-well microtiter plates or (25) boxes w/2 1/8" max height 12 1/8 18
£325.93GBP / Each
£391.12GBP (incl VAT)
COMPARE
(54 w/lids or 72 w/o lids) 96/384-well microtiter plates or (18) boxes w/2 1/8" max height 7 5/16 21 1/2
£235.08GBP / Each
£282.10GBP (incl VAT)
COMPARE
(72 w/lids or 96 w/o lids) 96/384-well microtiter plates or (24) boxes w/2 1/8" max height 9 11/16 21 1/2
£317.38GBP / Each
£380.86GBP (incl VAT)
COMPARE
(90 w/lids or 120 w/o lids) 96/384-well microtiter plates or (30) boxes w/2 1/8" max height 12 1/8 21 1/2
£369.52GBP / Each
£443.42GBP (incl VAT)
COMPARE
(108 w/lids or 144 w/o lids) 96/384-well microtiter plates or (36) boxes w/2 1/8" max height 14 1/2 21 1/2
£402.12GBP / Each
£482.54GBP (incl VAT)
COMPARE
(84 w/lids or 112 w/o lids) 96/384-well microtiter plates or (28) boxes w/2 1/8" max height 9 11/16 25
£339.38GBP / Each
£407.26GBP (incl VAT)
COMPARE
(105 w/lids or 140 w/o lids) 96/384-well microtiter plates or (35) boxes w/2 1/8" max height 12 1/8 25
£401.30GBP / Each
£481.56GBP (incl VAT)
Showing 1- 13 of 13