Cole-Parmer® Translucent Polypropylene Graduated Cylinders
Molded in one piece for enhanced durability
  • Wide hexagonal base provides stability
  • Accurate to ±2.5%
  • Molded in graduations will not wear off
  • Bulk packed for greater savings
Featuring pour spouts, molded in graduations, and heat resistance from 5 to 248°F (-15 to 150°C) these polypropylene cylinders are excellent for general use in the lab.
£15.27 - £42.73GBP / Pkg of 10
£18.32 - £51.28GBP (incl VAT)
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Description
Availability Pricing
COMPARE
50 1 Plastic Graduated Cylinder, Translucent PP, 50 mL, 10/pk
£15.27GBP / Pkg of 10
£18.32GBP (incl VAT)
COMPARE
100 1 Plastic Graduated Cylinder, Translucent PP, 100 mL, 10/pk
£21.41GBP / Pkg of 10
£25.69GBP (incl VAT)
COMPARE
250 5 Plastic Graduated Cylinder, Translucent PP, 250 mL, 10/pk
£42.73GBP / Pkg of 10
£51.28GBP (incl VAT)