Cole-Parmer® Threaded to Hose Barb Fitting, Natural Nylon
Threaded barb adapter design provides safe, difficult-to-break connection
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
Use these high-quality PVC-free fittings in biomedical research, pharmaceutical, electronic, and other general-purpose applications.
Quickly and easily transition from soft tubing to a system that accepts a threaded connection using threaded barb adapters. Threads provide a tight and secure connection; a single barb on the other end of the fitting creates a tight seal to keep the tubing in place. An anti-rotation device on the hose barb prevents tubing from rotating, minimizing kinks.
These natural nylon fittings provide good tensile strength, stiffness, and impact resistance and are resistant to fuels.
£9.65 - £15.35GBP / Pkg of 10
£11.58 - £18.42GBP (incl VAT)
18 variations of this product are available.
See All Product Options Below
18 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 2) (in) Barb Size / Tubing Id (Port 3) (in) Thread Size NPT(M) (Port 1) (InchShort)
Availability Pricing
COMPARE
1/8 1/8 1/8
£14.00GBP / Pkg of 10
£16.80GBP (incl VAT)
COMPARE
1/4 1/4 1/8
£9.65GBP / Pkg of 10
£11.58GBP (incl VAT)
COMPARE
3/8 3/8 1/8
£9.65GBP / Pkg of 10
£11.58GBP (incl VAT)
COMPARE
1/4 1/4 1/4
£9.65GBP / Pkg of 10
£11.58GBP (incl VAT)
COMPARE
3/8 3/8 1/4
£9.65GBP / Pkg of 10
£11.58GBP (incl VAT)
COMPARE
1/2 1/2 1/4
£9.85GBP / Pkg of 10
£11.82GBP (incl VAT)
COMPARE
1/4 1/4 3/8
£10.05GBP / Pkg of 10
£12.06GBP (incl VAT)
COMPARE
3/8 3/8 3/8
£10.05GBP / Pkg of 10
£12.06GBP (incl VAT)
COMPARE
1/2 1/2 3/8
£10.25GBP / Pkg of 10
£12.30GBP (incl VAT)
COMPARE
1/4 1/4 1/2
£10.80GBP / Pkg of 10
£12.96GBP (incl VAT)
COMPARE
3/8 3/8 1/2
£10.80GBP / Pkg of 10
£12.96GBP (incl VAT)
COMPARE
1/2 1/2 1/2
£10.80GBP / Pkg of 10
£12.96GBP (incl VAT)
COMPARE
3/4 3/4 1/2
£13.05GBP / Pkg of 10
£15.66GBP (incl VAT)
COMPARE
1/4 1/4 3/4
£13.05GBP / Pkg of 10
£15.66GBP (incl VAT)
COMPARE
3/8 3/8 3/4
£13.05GBP / Pkg of 10
£15.66GBP (incl VAT)
COMPARE
1/2 1/2 3/4
£13.05GBP / Pkg of 10
£15.66GBP (incl VAT)
COMPARE
1 1 3/4
£15.35GBP / Pkg of 10
£18.42GBP (incl VAT)
COMPARE
1 1 1
£15.35GBP / Pkg of 10
£18.42GBP (incl VAT)
Showing 1- 18 of 18