Cole-Parmer® Threaded to Hose Barb Fitting, HDPE
Easily transition from threaded connection to hose barbs
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
  • Single barb design tightly “grips” the tubing
  • Threaded barb adapter design provides safe, difficult-to-break connection
Use these high-quality PVC-free fittings in biomedical research, pharmaceutical, electronic, and other critical-use applications. Quickly and easily transition to and from soft tubing to a system that accepts a threaded connection using threaded barb adapters. Tee adapters feature one threaded port and two hose barbs to easily combine or split flow. Threads provide a tight and secure connection; a single barb on the other end of the fitting creates a tight seal to keep the tubing in place. An anti-rotation device on the hose barb prevents tubing from rotating, minimizing kinks. These high-density polyethylene (HDPE) fittings are softer than nylon, polypropylene, and Kynar®. They exhibit excellent resistance to chemicals but have lower thermal resistance than other materials.
£9.65 - £15.70GBP / Pkg of 10
£11.58 - £18.84GBP (incl VAT)
19 variations of this product are available.
See All Product Options Below
19 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 2) (in) Barb Size / Tubing Id (Port 3) (in) Thread Size NPT(M) (Port 1) (InchShort)
Availability Pricing
COMPARE
1/8 1/8 1/8
£15.70GBP / Pkg of 10
£18.84GBP (incl VAT)
COMPARE
1/4 1/4 1/8
£9.65GBP / Pkg of 10
£11.58GBP (incl VAT)
COMPARE
3/8 3/8 1/8
£9.65GBP / Pkg of 10
£11.58GBP (incl VAT)
COMPARE
1/4 1/4 1/4
£9.65GBP / Pkg of 10
£11.58GBP (incl VAT)
COMPARE
3/8 3/8 1/4
£9.65GBP / Pkg of 10
£11.58GBP (incl VAT)
COMPARE
1/2 1/2 1/4
£10.05GBP / Pkg of 10
£12.06GBP (incl VAT)
COMPARE
1/4 1/4 3/8
£10.05GBP / Pkg of 10
£12.06GBP (incl VAT)
COMPARE
3/8 3/8 3/8
£10.05GBP / Pkg of 10
£12.06GBP (incl VAT)
COMPARE
1/2 1/2 3/8
£10.25GBP / Pkg of 10
£12.30GBP (incl VAT)
COMPARE
1/4 1/4 1/2
£10.80GBP / Pkg of 10
£12.96GBP (incl VAT)
COMPARE
3/8 3/8 1/2
£10.80GBP / Pkg of 10
£12.96GBP (incl VAT)
COMPARE
1/2 1/2 1/2
£10.80GBP / Pkg of 10
£12.96GBP (incl VAT)
COMPARE
3/4 3/4 1/2
£12.90GBP / Pkg of 10
£15.48GBP (incl VAT)
COMPARE
1/4 1/4 3/4
£12.90GBP / Pkg of 10
£15.48GBP (incl VAT)
COMPARE
3/8 3/8 3/4
£12.90GBP / Pkg of 10
£15.48GBP (incl VAT)
COMPARE
1/2 1/2 3/4
£12.90GBP / Pkg of 10
£15.48GBP (incl VAT)
COMPARE
3/4 3/4 3/4
£12.90GBP / Pkg of 10
£15.48GBP (incl VAT)
COMPARE
1 1 3/4
£14.75GBP / Pkg of 10
£17.70GBP (incl VAT)
COMPARE
1 1 1
£14.75GBP / Pkg of 10
£17.70GBP (incl VAT)
Showing 1- 19 of 19