Cole-Parmer® Thermoplastic Vulcanizate (TPV) Pharma Grade Tubing
Offers great chemical and kink resistance for excellent pump performance
  • Biocompatible thermoplastic tubing offers good chemical resistance
  • Does not contain BPA, phthalates, PVC, or peroxides
  • Sterilize by autoclave, gamma irradiation, and chemical sterilants
  • Complies with U.S. Pharmacopeia 43, National Formulary 38 (USP), General Chapter <88> Class VI, Biological reactivity Tests, In Vivo (2020)
  • Ideal for general-purpose, laboratory, and single-use applications
  • Free technical application support available to assist with tubing selection
Use the biocompatible TPV tubing for peristaltic pumping, filling, and fluid transfer applications. This flexible opaque tubing offers excellent pump performance while providing great chemical resistance, making it a great alternative to silicone tubing.
TPV tubing does not contain BPA, phthalates, PVC, or peroxide that can leach out into the fluid, making it perfect for pharmaceutical, laboratory, and diagnostic applications. For an added level of particle control, a liner is placed between the tubing package and the box.
£152.39 - £424.33GBP / Pkg of 1
£182.87 - £509.20GBP (incl VAT)
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubing ID (in) Tubing OD (in) Wall Thickness (in)
Availability Pricing
COMPARE
1/16 3/16 1/16
£152.39GBP / Pkg of 1
£182.87GBP (incl VAT)
COMPARE
1/8 1/4 1/16
£175.83GBP / Pkg of 1
£211.00GBP (incl VAT)
COMPARE
3/16 5/16 1/16
£181.30GBP / Pkg of 1
£217.56GBP (incl VAT)
COMPARE
1/4 1/2 1/8
£361.81GBP / Pkg of 1
£434.17GBP (incl VAT)
COMPARE
3/8 5/8 1/8
£355.56GBP / Pkg of 1
£426.67GBP (incl VAT)
COMPARE
3/16 7/16 1/8
£195.36GBP / Pkg of 1
£234.43GBP (incl VAT)
COMPARE
1/4 3/8 1/16
£212.56GBP / Pkg of 1
£255.07GBP (incl VAT)
COMPARE
1/4 7/16 3/32
£275.85GBP / Pkg of 1
£331.02GBP (incl VAT)
COMPARE
1/2 3/4 1/8
£424.33GBP / Pkg of 1
£509.20GBP (incl VAT)
Showing 1- 9 of 9