Cole-Parmer® Thermoplastic (SEBS) Tubing
Easily weldable/sealable—compatible with all common tube sealers and aseptic tube welders
  • Specifically designed for demanding biopharmaceutical applications
  • Does not contain BPA, phthalates, PVC, or peroxides
  • Sterilize by autoclave, gamma irradiation, and chemical sterilants
  • Complies with U.S. Pharmacopeia 43, National Formulary 38 (USP), General Chapter <88> Class VI, Biological reactivity Tests, In Vivo (2020)
  • Ideal for general-purpose, laboratory, and single-use applications
  • Free technical application support available to assist with tubing selection
Designed for the most demanding biopharmaceutical applications, the thermoplastic SEBS (styrene-ethylene-butylene-styrene) tubing exhibits superior gas barrier properties and excellent pressure performance. Use the tubing in peristaltic pumps, single-use assemblies, process filling, and sampling devices.
Thermoplastic (SEBS) tubing does not contain BPA, phthalates, PVC, or peroxides that can leach out into the fluid, making it ideal for pharmaceutical applications, buffer and media preparation, and cell media processing. For an added level of particle control, a liner is placed between the tubing package and the box.
£58.75 - £292.60GBP / Pkg of 1
£70.50 - £351.12GBP (incl VAT)
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubing ID (in) Tubing OD (in) Wall Thickness (in)
Availability Pricing
COMPARE
1/16 3/16 1/16
£58.75GBP / Pkg of 1
£70.50GBP (incl VAT)
COMPARE
1/8 1/4 1/16
£75.93GBP / Pkg of 1
£91.12GBP (incl VAT)
COMPARE
3/16 5/16 1/16
£88.47GBP / Pkg of 1
£106.16GBP (incl VAT)
COMPARE
1/4 1/2 1/8
£171.15GBP / Pkg of 1
£205.38GBP (incl VAT)
COMPARE
3/8 5/8 1/8
£231.00GBP / Pkg of 1
£277.20GBP (incl VAT)
COMPARE
3/16 7/16 1/8
£74.55GBP / Pkg of 1
£89.46GBP (incl VAT)
COMPARE
1/4 3/8 1/16
£87.23GBP / Pkg of 1
£104.68GBP (incl VAT)
COMPARE
1/4 7/16 3/32
£129.15GBP / Pkg of 1
£154.98GBP (incl VAT)
COMPARE
1/2 3/4 1/8
£292.60GBP / Pkg of 1
£351.12GBP (incl VAT)
Showing 1- 9 of 9