Cole-Parmer® Support Plates
Provide support for your specific laboratory equipment
Use to support your labware items from lab fames or support stands. The round plate design features a raised rim to prevent your apparatus from falling, requires an additional connector in order to attach clamps to frames or other apparatus. The rectangular plates include an integral connector.
£94.17 - £338.08GBP / Each
£113.00 - £405.70GBP (incl VAT)
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Round Support Plate, 6" Dia Plate with 1/2" Dia Rod, Aluminum
£94.17GBP / Each
£113.00GBP (incl VAT)
COMPARE
Frame Rod Platform, Aluminum, 9-1/2" L x 6-1/2" W
£169.87GBP / Each
£203.84GBP (incl VAT)
COMPARE
Frame Rod Platform, Aluminum, 12-3/4" L x 9" W
£269.63GBP / Each
£323.56GBP (incl VAT)
COMPARE
Frame Rod Platform, Aluminum, 15-3/4" L x 12" W
£338.08GBP / Each
£405.70GBP (incl VAT)