Cole-Parmer® Sterile WFI Samplers
A fused sanitary fitting with built-in gasket offers contamination-free sample collection
  • One-piece design with sanitary fitting provides clean, hassle-free connection
  • Gamma irradiated for sterility
  • Double-bagged in an easy-peel, low-permeability pouch
  • Assembled in ISO Class 7 cleanroom
  • USP Class VI, REACH, and RoHS compliant
Minimize the risk of sample contamination using the sterile WFI samplers. Samplers feature a polypropylene (PP) sanitary fitting with a fused PP gasket for sample collection without contact with the fitting, removing an immense contamination risk. Samplers come gamma irradiated for microbial control and double-bagged in an easy-peel pouch.
£541.49 - £612.60GBP / Pkg of 10
£649.79 - £735.12GBP (incl VAT)
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
End Fitting Length (InchShort)
Availability Pricing
COMPARE
3/4" Tri-Clamp 18
£541.49GBP / Pkg of 10
£649.79GBP (incl VAT)
COMPARE
3/4" Tri-Clamp 36
£612.60GBP / Pkg of 10
£735.12GBP (incl VAT)
COMPARE
1-1/2" Tri-Clamp 18
£541.49GBP / Pkg of 10
£649.79GBP (incl VAT)
COMPARE
1-1/2" Tri-Clamp 36
£612.60GBP / Pkg of 10
£735.12GBP (incl VAT)
COMPARE
3/4" Tri-Clamp 18
£541.49GBP / Pkg of 10
£649.79GBP (incl VAT)
COMPARE
3/4" Tri-Clamp 36
£612.60GBP / Pkg of 10
£735.12GBP (incl VAT)
COMPARE
1-1/2" Tri-Clamp 18
£541.49GBP / Pkg of 10
£649.79GBP (incl VAT)
COMPARE
1-1/2" Tri-Clamp 36
£612.60GBP / Pkg of 10
£735.12GBP (incl VAT)
Showing 1- 8 of 8