Cole-Parmer® Sterile Volumetric Lab Sampling Spoons, HDPE
Accurately measures set volumes
  • Gamma irradiation sterilized (SAL 10-6)
  • Manufactured in an ISO 8 (Class 100,000) cleanroom to ensure cleanliness
  • High-quality HDPE conforms to FDA, EC, and EU regulations
  • BSE/TSE free
Reduce cross contamination when sampling powders, granules, liquids, gels, and suspensions with these sterile, disposable lab spoons. Designed to enable set volumes to be accurately measured, these lab sampling spoons are gamma irradiation sterilized (SAL 10-6) and individually packaged in a heat-sealed PE/PET bag so they can be used straight out of the bag without cleaning. Handles measure about 13.6 cm (5.4 in) long to help keep hands away from the materials being measured.
Molded in an ISO 8 (Class 100,000) clean room, these laboratory spoons are FDA CFR 177.1520, EU regulation 10/2011 compliant, EC regulation 1935/2004 compliant, and BSE/TSE free.
£135.03 - £150.58GBP / Pkg of 100
£162.04 - £180.70GBP (incl VAT)
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Overall Length (cm) Width (cm)
Availability Pricing
COMPARE
1 15.3 1.8
£135.03GBP / Pkg of 100
£162.04GBP (incl VAT)
COMPARE
2.5 16 2.3
£137.72GBP / Pkg of 100
£165.26GBP (incl VAT)
COMPARE
5 16.5 2.9
£135.03GBP / Pkg of 100
£162.04GBP (incl VAT)
COMPARE
10 17.1 3.5
£135.03GBP / Pkg of 100
£162.04GBP (incl VAT)
COMPARE
20 18 4.3
£135.03GBP / Pkg of 100
£162.04GBP (incl VAT)
COMPARE
30 18.7 5
£150.58GBP / Pkg of 100
£180.70GBP (incl VAT)